1 - 30 of 30 products displayed
  • Phospho-IL10R / IL-10 Receptor alpha pTyr496 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA535565
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Interleukin-10 receptor subunit 1; CDw210a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535565 Not Orderable Online
Check your price
  • IL20RA Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141167
  • Size 100µg
Applications
Flow Cytometry (2-5 µg/10^6 cells), Western Blot (1-3 µg/ml)
Alternate names:
ZcytoR7; IL-20R1; Cytokine receptor family 2 member 8; Cytokine receptor class-II member 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141167 375.00
Check your price
  • CD123 Monoclonal Antibody, PerCP conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PerCP
  • Catalog # MA523601
  • Size 100Tests
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Interleukin-3 receptor class II alpha chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523601 289.00
Check your price
  • Phospho-IL10RA (Ser319, Ser323) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537608
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Interleukin-10 receptor subunit 1; CDw210a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537608 309.00
Check your price
  • CD123 Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523818
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunohistochemistry (8-25 µg/ml), Neutralization (0.6-1.2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
CD123, IL3R, IL3RAY, IL3RX, IL3RY, hIL-3Ra, CD123 antigen, IL-3 receptor alpha SP2 isoform, IL-3 receptor subunit alpha, IL-3R subunit alpha, IL-3R-alpha, IL-3RA, interleukin-3 receptor subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523818 275.00
Check your price
  • CD123 Monoclonal Antibody, PerCP-Cyanine5.5 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PerCP-Cyanine5.5
  • Catalog # A16426
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD123, IL3R, IL3RAY, IL3RX, IL3RY, hIL-3Ra, CD123 antigen, IL-3 receptor alpha SP2 isoform, IL-3 receptor subunit alpha, IL-3R subunit alpha, IL-3R-alpha, IL-3RA, interleukin-3 receptor subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16426 138.00
Check your price
  • CD123 Monoclonal Antibody, PE-Cyanine7 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE-Cyanine7
  • Catalog # A16422
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD123, IL3R, IL3RAY, IL3RX, IL3RY, hIL-3Ra, CD123 antigen, IL-3 receptor alpha SP2 isoform, IL-3 receptor subunit alpha, IL-3R subunit alpha, IL-3R-alpha, IL-3RA, interleukin-3 receptor subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16422 158.00
Check your price
  • CD123 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # A16377
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD123, IL3R, IL3RAY, IL3RX, IL3RY, hIL-3Ra, CD123 antigen, IL-3 receptor alpha SP2 isoform, IL-3 receptor subunit alpha, IL-3R subunit alpha, IL-3R-alpha, IL-3RA, interleukin-3 receptor subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16377 109.00
Check your price
  • CD123 Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # A16417
  • Size 100tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD123, IL3R, IL3RAY, IL3RX, IL3RY, hIL-3Ra, CD123 antigen, IL-3 receptor alpha SP2 isoform, IL-3 receptor subunit alpha, IL-3R subunit alpha, IL-3R-alpha, IL-3RA, interleukin-3 receptor subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16417 268.00
Check your price
  • CD123 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A16399
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD123, IL3R, IL3RAY, IL3RX, IL3RY, hIL-3Ra, CD123 antigen, IL-3 receptor alpha SP2 isoform, IL-3 receptor subunit alpha, IL-3R subunit alpha, IL-3R-alpha, IL-3RA, interleukin-3 receptor subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16399 85.75
Check your price
  • IL4R Monoclonal Antibody for Western Blot, IHC, Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523814
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (8-25 µg/ml), Neutralization (3-6 ng/ml), Western Blot (1 µg/ml)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523814 275.00
Check your price
  • IL22RA1 Monoclonal Antibody for IF, ICC and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524205
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
CRF2-9, IL22R, IL22R1, IL-22 receptor subunit alpha-1, IL-22R-alpha-1, IL-22RA1, cytokine receptor class-II member 9, cytokine receptor family 2 member 9, interleukin-22 receptor subunit alpha-1, zcytoR11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524205 275.00
Check your price
  • IL4R Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536394
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536394 309.00
Check your price
  • IL4R Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547297
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547297 349.00
Check your price
  • IL15RA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534512
  • Size 100ug
Applications
Western Blot (1-2 ug/mL)
Alternate names:
IL-15RA, Interleukin-15 receptor subunit alpha, IL15RA, CD215
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534512 349.00
Check your price
  • IL10RA Polyclonal Antibody for Western Blot, IF, ICC and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547383
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (20-40 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-10 receptor subunit 1; CDw210a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547383 349.00
Check your price
  • IL10RA Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527772
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Interleukin-10 receptor subunit 1; CDw210a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527772 349.00
Check your price
  • IL31RA Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547391
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
ZcytoR17; Gp130-like monocyte receptor; GLM-R; Cytokine receptor-like 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547391 349.00
Check your price
  • IL6RA Monoclonal Antibody for Western Blot, IF and ICC
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523951
  • Size 100µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
IL-6R 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523951 275.00
Check your price
  • Il13ra2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547732
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL13R
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547732 349.00
Check your price
  • IL22RA1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519987
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1 µg/ml)
Alternate names:
ZcytoR11; Cytokine receptor family 2 member 9; Cytokine receptor class-II member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519987 349.00
Check your price
  • IL22RA1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519986
  • Size 100ug
Applications
Western Blot (1 µg/ml)
Alternate names:
ZcytoR11; Cytokine receptor family 2 member 9; Cytokine receptor class-II member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519986 349.00
Check your price
  • IL-27 Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546993
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p28; Interleukin-30; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546993 375.00
Check your price
  • CD25 Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M10AR
  • Size 100tests
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p55; TAC antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
M10AR 275.00
Check your price
  • CD123 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523460
  • Size 100ug
Applications
Western Blot (3-5 µg/ml)
Alternate names:
CD123, IL3R, IL3RAY, IL3RX, IL3RY, hIL-3Ra, CD123 antigen, IL-3 receptor alpha SP2 isoform, IL-3 receptor subunit alpha, IL-3R subunit alpha, IL-3R-alpha, IL-3RA, interleukin-3 receptor subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523460 349.00
Check your price
  • CD3/CD25 Monoclonal Antibody, FITC, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112091
  • Size 100Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; p55; TAC antigen; Growth arrest-specific protein 3; T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112091 375.00
Check your price
  • CD3/CD25 Monoclonal Antibody, FITC, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112092
  • Size 50Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; p55; TAC antigen; Growth arrest-specific protein 3; T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112092 309.00
Check your price
  • TCCR Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519984
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
ZcytoR1; Type I T-cell cytokine receptor; Cytokine receptor-like 1; Cytokine receptor WSX-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519984 349.00
Check your price
  • TCCR Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519985
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
ZcytoR1; Type I T-cell cytokine receptor; Cytokine receptor-like 1; Cytokine receptor WSX-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519985 349.00
Check your price
  • IL13RA2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA544066
  • Size 100µl
Applications
Western Blot (1:1000)
Alternate names:
Interleukin-13-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544066 309.00
Check your price