1 - 30 of 30 products displayed
  • IL-6 Receptor Monoclonal Antibody for IF, IHC (F), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180456
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1:50), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Membrane glycoprotein 80; IL-6R 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180456 319.00
Check your price
  • IL-6 Receptor Monoclonal Antibody for IHC, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHR0961
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Membrane glycoprotein 80; IL-6R 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHR0961 304.00
Check your price
  • IL-6 Receptor Polyclonal Antibody for Western Blot, IHC (F), Flow, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547209
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.2-1 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL-6R 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547209 349.00
Check your price
  • IL4R Monoclonal Antibody for Western Blot, IHC, Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523814
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (8-25 µg/ml), Neutralization (3-6 ng/ml), Western Blot (1 µg/ml)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523814 275.00
Check your price
  • IL4R Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547297
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547297 349.00
Check your price
  • IL4R Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536394
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536394 309.00
Check your price
  • Phospho-IL4R (Tyr497) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538614
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538614 309.00
Check your price
  • IL4R Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538615
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538615 309.00
Check your price
  • IL4R Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA536522
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536522 309.00
Check your price
  • CD25 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Mammal
  • Conjugate FITC
  • Catalog # MA135144
  • Size 500 µL
Applications
Flow Cytometry (5 µl per 1x10^6 cells.)
Alternate names:
p55; TAC antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135144 425.00
Check your price
  • CD25 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Mammal
  • Conjugate PE
  • Catalog # MA135145
  • Size 500 µL
Applications
Flow Cytometry (5 µl per 1x10^6 cells.)
Alternate names:
p55; TAC antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135145 550.00
Check your price
  • IL20RA Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141167
  • Size 100 µg
Applications
Flow Cytometry (2-5 µg/10^6 cells), Western Blot (1-3 µg/ml)
Alternate names:
ZcytoR7; IL-20R1; Cytokine receptor family 2 member 8; Cytokine receptor class-II member 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141167 375.00
Check your price
  • TCCR Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523965
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
WSX-1; Type I T-cell cytokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523965 275.00
Check your price
  • TCCR Monoclonal Antibody for Western Blot
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523966
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
WSX-1; Type I T-cell cytokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523966 275.00
Check your price
  • CD127 Monoclonal Antibody, Alexa Fluor® 488 conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # A14809
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Interleukin-7 Receptor alpha, CD127
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14809 140.00
Check your price
  • CD127 Monoclonal Antibody, PerCP-Cyanine5.5 conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PerCP-Cyanine5.5
  • Catalog # A14799
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Interleukin-7 Receptor alpha, CD127
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14799 134.00
Check your price
  • CD127 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A14759
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Interleukin-7 Receptor alpha, CD127
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14759 116.00
Check your price
  • Phospho-IL13RA1 (Tyr405) Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538607
  • Size 100 µg
Applications
Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Novel cytokine receptor 4; Interleukin-13-binding protein; Cancer/testis antigen 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538607 309.00
Check your price
  • TCCR Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA548004
  • Size 100 µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
WSX-1; Type I T-cell cytokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548004 309.00
Check your price
  • IL13RA1 Monoclonal Antibody for IF, ICC, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117751
  • Size 100 µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 ug/10^6 cells), Immunocytochemistry (1ug/10^6 cells), Immunofluorescence (1 ug/10^6 cells)
Alternate names:
Cancer/testis antigen 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117751 550.00
Check your price
  • IL13RA1 Monoclonal Antibody for Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117750
  • Size 100 µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 ug/10^6 cells)
Alternate names:
Cancer/testis antigen 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117750 550.00
Check your price
  • IL13RA1 Polyclonal Antibody for Western Blot, IHC (P), Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547149
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (10-30 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cancer/testis antigen 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547149 349.00
Check your price
  • IL13RA2 Polyclonal Antibody for Western Blot, IHC (P), Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546976
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.2-10 µg/ml)
Alternate names:
Interleukin-13-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546976 375.00
Check your price
  • Phospho-Il13Ra1 (Tyr405) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA540237
  • Size 100 µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Novel cytokine receptor 4; Interleukin-13-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540237 309.00
Check your price
  • Il13ra2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547732
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL13R
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547732 349.00
Check your price
  • CD25 Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M10AR
  • Size 100 tests
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p55; TAC antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
M10AR 275.00
Check your price
  • IL-27 p28 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141055
  • Size 100 µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
p28; Interleukin-30
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141055 309.00
Check your price
  • IL22RA2 Polyclonal Antibody for IF, ICC, IHC (P), Flow and ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121359
  • Size 100 µg
Applications
ELISA (1:100,000), Flow Cytometry (Assay dependent), Immunocytochemistry (1:300), Immunofluorescence (1:300), Immunohistochemistry (Paraffin) (Assay-dependent)
Alternate names:
ZcytoR16; Interleukin-22-binding protein; Cytokine receptor family type 2, soluble 1; Cytokine receptor family 2 member 10; Cytokine receptor class-II member 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121359 425.00
Check your price
  • IL-2R alpha / CD25 Monoclonal Antibody for IHC (F), Flow
  • Host Mouse
  • Target Species Human
  • Conjugate
  • Catalog # MA191221
  • Size 500µL
Applications
Flow Cytometry (1:150-1:300), Immunohistochemistry (Frozen) (1:50-1:100)
Alternate names:
p55; TAC antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191221 369.00
Check your price
  • IL10RA Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA136187
  • Size 100 µg
Applications
Flow Cytometry (1:10)
Alternate names:
Interleukin-10 receptor subunit 1; CDw210a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA136187 700.00
Check your price