1 - 30 of 30 products displayed
  • IL-6 Receptor Polyclonal Antibody for Western Blot, IHC (F), Flow, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547209
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.2-1 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL-6R 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547209 349.00
Check your price
  • IL-6 Receptor Monoclonal Antibody for Western Blot, IF and ICC
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523951
  • Size 100 µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
IL-6R 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523951 275.00
Check your price
  • CD25 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Mammal
  • Conjugate FITC
  • Catalog # MA135144
  • Size 500 µL
Applications
Flow Cytometry (5 µl per 1x10^6 cells.)
Alternate names:
p55; TAC antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135144 425.00
Check your price
  • IL5RA Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547750
  • Size 100 µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
CD125, CDw125, HSIL5R3, IL5R, CD125 antigen, IL-5 receptor subunit alpha, IL-5R subunit alpha, interleukin 5 receptor type 3, interleukin-5 receptor alpha chain, interleukin-5 receptor subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547750 349.00
Check your price
  • CD127 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547927
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-7 Receptor alpha, CD127
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547927 349.00
Check your price
  • CD25 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Mammal
  • Conjugate PE
  • Catalog # MA135145
  • Size 500 µL
Applications
Flow Cytometry (5 µl per 1x10^6 cells.)
Alternate names:
p55; TAC antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135145 550.00
Check your price
  • IL1R1 Polyclonal Antibody for Western Blot, IHC and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547937
  • Size 100 µg
Applications
Immunohistochemistry (5-15 µg/ml), Neutralization (0.2-1.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
p80; Interleukin-1 receptor type I; Interleukin-1 receptor alpha; CD121 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547937 349.00
Check your price
  • IL22RA1 Monoclonal Antibody for IF, ICC and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524205
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
CRF2-9, IL22R, IL22R1, IL-22 receptor subunit alpha-1, IL-22R-alpha-1, IL-22RA1, cytokine receptor class-II member 9, cytokine receptor family 2 member 9, interleukin-22 receptor subunit alpha-1, zcytoR11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524205 275.00
Check your price
  • IL4R Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547297
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547297 349.00
Check your price
  • IL5RA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527900
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527900 349.00
Check your price
  • CD127 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523825
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
CDw127
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523825 275.00
Check your price
  • CD127 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547443
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
CDw127
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547443 349.00
Check your price
  • IL11RA Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547667
  • Size 100 µg
Applications
Neutralization (2-6 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Novel cytokine receptor 1; Enhancer trap locus homolog 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547667 349.00
Check your price
  • IL5RA (soluble) Polyclonal Antibody for Western Blot, Flow, ELISA, Neu and IA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124792
  • Size 25 µg
Applications
ELISA (0.5-1 µg/ml), Flow Cytometry (50-250 ng for 10^6 cells), Inhibition Assays (1-3 µg/ml), Neutralization (1-3 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124792 375.00
Check your price
  • IL-12 p35 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523226
  • Size 100 ug
Applications
Western Blot (3-5 µg/ml)
Alternate names:
NK cell stimulatory factor chain 1; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523226 349.00
Check your price
  • IL4R Monoclonal Antibody for Western Blot, IHC, Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523814
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (8-25 µg/ml), Neutralization (3-6 ng/ml), Western Blot (1 µg/ml)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523814 275.00
Check your price
  • IL10RA Polyclonal Antibody for Western Blot, IF, ICC and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547383
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (20-40 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-10 receptor subunit 1; CDw210a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547383 349.00
Check your price
  • IL-12 p70 Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523715
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
NK cell stimulatory factor chain 1; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523715 349.00
Check your price
  • CD122 Polyclonal Antibody for Western Blot and Flow
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547622
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
p70-75; High affinity IL-2 receptor subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547622 349.00
Check your price
  • IL1R2 Polyclonal Antibody, Alexa Fluor® 488 conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # PA546897
  • Size 100 Tests
Applications
Flow Cytometry (5 µl/10^6 cells)
Alternate names:
Interleukin-1 receptor type II; Interleukin-1 receptor beta; IL-1 type II receptor; CD121 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546897 289.00
Check your price
  • IL1R2 Polyclonal Antibody for Western Blot, IHC (F) and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547759
  • Size 100 µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-1 receptor type II; Interleukin-1 receptor beta; IL-1 type II receptor; CD121 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547759 349.00
Check your price
  • IL5RA Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523621
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523621 309.00
Check your price
  • IL-12 p35 Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA175190
  • Size 500 µg
Applications
ELISA (Assay Dependent)
Alternate names:
NK cell stimulatory factor chain 1; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175190 1,500.00
Check your price
  • IL5RA Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523581
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523581 279.00
Check your price
  • IL5RA Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547340
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (1-3 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547340 349.00
Check your price
  • CD45 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517959
  • Size 100 ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517959 129.00
Check your price
  • CD45 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517960
  • Size 300 ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517960 309.00
Check your price
  • CD45 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517961
  • Size 100 ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517961 139.00
Check your price
  • CD45 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517962
  • Size 300 ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517962 309.00
Check your price
  • CD45 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517963
  • Size 50 ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517963 129.00
Check your price