1 - 27 of 27 products displayed
  • CD127 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547927
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-7 Receptor alpha, CD127
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547927 349.00
Check your price
  • CD127 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547443
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
CDw127
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547443 349.00
Check your price
  • CD127 Antibody (Monoclonal, 40131)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523825
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
CDw127
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523825 275.00
Check your price
  • Phospho-CD127 (Tyr449) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538619
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
CDw127
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538619 309.00
Check your price
  • IL10RA Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527772
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Interleukin-10 receptor subunit 1; CDw210a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527772 349.00
Check your price
  • IL22RA2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521490
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:100-1:1000)
Alternate names:
ZcytoR16; Interleukin-22-binding protein; Cytokine receptor family type 2, soluble 1; Cytokine receptor family 2 member 10; Cytokine receptor class-II member 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521490 349.00
Check your price
  • IL13RA1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528309
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Cancer/testis antigen 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528309 349.00
Check your price
  • IL13RA1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530071
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Cancer/testis antigen 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530071 349.00
Check your price
  • CD127 Antibody, PE
  • Host Sheep
  • Target Species Rat
  • Conjugate PE
  • Catalog # PA546900
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Interleukin-7 Receptor alpha, CD127
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546900 339.00
Check your price
  • CD127 Antibody
  • Host Sheep
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547758
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Interleukin-7 Receptor alpha, CD127
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547758 349.00
Check your price
  • IL-12/IL-23 p40 Antibody, Biotin (Monoclonal, C8.6)
  • Host Rat
  • Target Species Human
  • Conjugate Biotin See Additional Formats Unconjugated
  • Catalog # M121B
  • Size 250 ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
NK cell stimulatory factor chain 1; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
M121B 416.00
Check your price
  • IL10RA Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535361
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Interleukin-10 receptor subunit 1; CDw210a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535361 349.00
Check your price
  • IL-12 p70 Antibody (Monoclonal, 20C2)
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M122
  • Size 500 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
NK cell stimulatory factor chain 1; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
M122 375.00
Check your price
  • IL2RG Antibody (Monoclonal, 633162)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524286
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (0.08-0.4 µg/ml)
Alternate names:
p64; gammaC; Interleukin-2 receptor subunit gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524286 275.00
Check your price
  • IL2RG Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547409
  • Size 250 µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Neutralization (20-60 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
p64; gammaC; Interleukin-2 receptor subunit gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547409 349.00
Check your price
  • IL5RA Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527900
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527900 349.00
Check your price
  • IL5RA Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547340
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (1-3 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547340 349.00
Check your price
  • CD130 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528932
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Oncostatin-M receptor subunit alpha; Membrane glycoprotein 130; Interleukin-6 signal transducer; CDw130
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528932 349.00
Check your price
  • CD122 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528835
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p70-75; High affinity IL-2 receptor subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528835 349.00
Check your price
  • CD122 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529818
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
p70-75; High affinity IL-2 receptor subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529818 349.00
Check your price
  • IL-15 Antibody (Monoclonal, 34505)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523729
  • Size 500 µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.1-0.2 µg/ml)
Alternate names:
IL-15, Interleukin-15, IL15
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523729 349.00
Check your price
  • IL-12/IL-23 p40 Antibody (Monoclonal, 31052)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523724
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture))
Alternate names:
NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523724 349.00
Check your price
  • IL2RG Antibody (Monoclonal, 644346)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524290
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
p64; gammaC; Interleukin-2 receptor subunit gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524290 275.00
Check your price
  • IL2RG Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547944
  • Size 100 µg
Applications
Neutralization (6-24 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
p64; gammaC; Interleukin-2 receptor subunit gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547944 349.00
Check your price
  • Integrin beta-1 / CD29 Antibody Monoclonal, Cy® 7, PE conjugate (HM-beta-1-1) (A14889)
  • Host Hamster
  • Target Species Mouse, Rat
  • Conjugate
  • Catalog # A14889
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-4 subunit beta; Fibronectin receptor subunit beta; Beta oligodendroglia
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14889 121.00
Check your price
  • Phospho-ITGB1 (Ser785) Antibody
  • Host Rabbit
  • Target Species Chicken, Mouse
  • Conjugate Unconjugated
  • Catalog # 44870G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
VLA-4 subunit beta; Fibronectin receptor subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
44870G 309.00
Check your price
  • NFATC2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534940
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
T-cell transcription factor NFAT1; NFAT pre-existing subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534940 349.00
Check your price