1 - 30 of 30 products displayed
  • IL-22 Receptor alpha Antibody (Monoclonal, 496514)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524205
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
CRF2-9, IL22R, IL22R1, IL-22 receptor subunit alpha-1, IL-22R-alpha-1, IL-22RA1, cytokine receptor class-II member 9, cytokine receptor family 2 member 9, interleukin-22 receptor subunit alpha-1, zcytoR11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524205 275.00
Check your price
  • IL-6 Receptor Antibody (Monoclonal, 255820)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523951
  • Size 100 µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
IL-6R 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523951 275.00
Check your price
  • Phospho-IL-8R beta / CXCR2 pSer347 Antibody Polyclonal (PA5-35693)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate
  • Catalog # PA535693
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
High affinity interleukin-8 receptor B; GRO/MGSA receptor; IL-8 receptor type 2; CDw128b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535693 339.00
Check your price
  • IL-22 Receptor alpha Antibody (Monoclonal, 305405)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524017
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
ZcytoR11; Cytokine receptor family 2 member 9; Cytokine receptor class-II member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524017 275.00
Check your price
  • IL-22 Receptor alpha Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519987
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1 µg/ml)
Alternate names:
ZcytoR11; Cytokine receptor family 2 member 9; Cytokine receptor class-II member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519987 349.00
Check your price
  • IL-22 Receptor alpha Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519986
  • Size 100 ug
Applications
Western Blot (1 µg/ml)
Alternate names:
ZcytoR11; Cytokine receptor family 2 member 9; Cytokine receptor class-II member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519986 349.00
Check your price
  • Phospho-CXCR2 (Ser347) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA538620
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
High affinity interleukin-8 receptor B; GRO/MGSA receptor; IL-8 receptor type 2; CDw128b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538620 309.00
Check your price
  • CXCR2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131217
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (1:250), Western Blot (1:500)
Alternate names:
High affinity interleukin-8 receptor B; GRO/MGSA receptor; IL-8 receptor type 2; CDw128b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131217 499.00
Check your price
  • IL20RA Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141167
  • Size 100 µg
Applications
Flow Cytometry (2-5 µg/10^6 cells), Western Blot (1-3 µg/ml)
Alternate names:
ZcytoR7; IL-20R1; Cytokine receptor family 2 member 8; Cytokine receptor class-II member 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141167 375.00
Check your price
  • IL-27 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520241
  • Size 100 ug
Applications
Western Blot (2-4 ug/ml)
Alternate names:
Epstein-Barr virus-induced gene 3 protein homolog; p28; Interleukin-30; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520241 349.00
Check your price
  • IL-27 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520240
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (2-4 ug/ml)
Alternate names:
Epstein-Barr virus-induced gene 3 protein homolog; p28; Interleukin-30; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520240 349.00
Check your price
  • IL-23 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520239
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520239 349.00
Check your price
  • IL-23 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520238
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520238 349.00
Check your price
  • IL-23 Antibody (Monoclonal, 320234)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523793
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523793 349.00
Check your price
  • IL-23 Antibody (Oligoclonal, 26HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710259
  • Size 100 µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
710259 309.00
Check your price
  • IL-27 p28 Antibody (Oligoclonal, 7HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710175
  • Size 100 µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
p28; Interleukin-30
Catalog # Size Price (AUD) Your Price (AUD) Qty
710175 309.00
Check your price
  • IL-27 p28 Antibody (Monoclonal, 7H7L10)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701073
  • Size 100 µg
Applications
Western Blot (0.1-1 ug/ml)
Alternate names:
p28; Interleukin-30
Catalog # Size Price (AUD) Your Price (AUD) Qty
701073 309.00
Check your price
  • IL-12 p70 Antibody, Biotin (Monoclonal, C17-8)
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MM121B
  • Size 250 ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MM121B 375.00
Check your price
  • IL-12 p70 Antibody (Monoclonal, C17-8)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MM121
  • Size 500 ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MM121 349.00
Check your price
  • IL20RA Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547086
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
ZcytoR7; IL-20R1; Cytokine receptor family 2 member 8; Cytokine receptor class-II member 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547086 349.00
Check your price
  • CXCR1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533453
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (15-20 µg/ml)
Alternate names:
IL-8 receptor type 1; High affinity interleukin-8 receptor A; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533453 349.00
Check your price
  • CXCR1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533454
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (16 µg/ml)
Alternate names:
IL-8 receptor type 1; High affinity interleukin-8 receptor A; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533454 349.00
Check your price
  • CXCR1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533452
  • Size 50 ug
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (7-11 µg/ml)
Alternate names:
IL-8 receptor type 1; High affinity interleukin-8 receptor A; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533452 349.00
Check your price
  • CXCR1 / IL8RA Antibody Monoclonal (501) (MA1-12878)
  • Host Mouse
  • Target Species Human
  • Conjugate
  • Catalog # MA112878
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
IL-8 receptor type 1; High affinity interleukin-8 receptor A; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112878 369.00
Check your price
  • CXCR1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141096
  • Size 100 µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
IL-8 receptor type 1; High affinity interleukin-8 receptor A; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141096 375.00
Check your price
  • CXCR1 / IL8RA Antibody Monoclonal (501) (AHR1522X)
  • Host Mouse
  • Target Species Human
  • Conjugate
  • Catalog # AHR1522X
  • Size 500µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
IL-8 receptor type 1; High affinity interleukin-8 receptor A; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHR1522X 419.00
Check your price
  • CXCR1 Antibody (Monoclonal, 42705.111)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124668
  • Size 100 µg
Applications
Flow Cytometry (10 µl for 10^6 PBMC), Inhibition Assays (0.1-0.3 µg/ml)
Alternate names:
IL-8 receptor type 1; High affinity interleukin-8 receptor A; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124668 375.00
Check your price
  • CXCR1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rabbit
  • Conjugate Unconjugated
  • Catalog # PA129466
  • Size 100 µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
High affinity interleukin-8 receptor A; IL-8 receptor type 1; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129466 425.00
Check your price
  • CXCR1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527184
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
IL-8 receptor type 1; High affinity interleukin-8 receptor A; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527184 349.00
Check your price
  • CXCR1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132560
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
IL-8 receptor type 1; High affinity interleukin-8 receptor A; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132560 399.00
Check your price