1 - 30 of 30 products displayed
  • IL-4 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA525165
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Lymphocyte stimulatory factor 1; IGG1 induction factor; B-cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; Pitrakinra; Binetrakin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525165 349.00
Check your price
  • IL-4 Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MM450C
  • Size 500µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Lymphocyte stimulatory factor 1; IGG1 induction factor; B-cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MM450C 349.00
Check your price
  • IL-4 Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M2IL4BI
  • Size 100ug
Applications
ELISA (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
M2IL4BI 349.00
Check your price
  • IL-4 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M3IL4I
  • Size 500ug
Applications
ELISA (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
M3IL4I 349.00
Check your price
  • IL-4 Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Rat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M450B
  • Size 250ug
Applications
Western Blot (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
M450B 349.00
Check your price
  • IL-4 Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M450
  • Size 500ug
Applications
Western Blot (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
M450 349.00
Check your price
  • IL4R Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536394
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536394 309.00
Check your price
  • IL4R Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA536522
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536522 309.00
Check your price
  • IL-4 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523722
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.1-0.6 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Lymphocyte stimulatory factor 1; IGG1 induction factor; B-cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523722 349.00
Check your price
  • IL-4 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 710254
  • Size 100µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Lymphocyte stimulatory factor 1; IGG1 induction factor; B-cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
710254 309.00
Check your price
  • IL-4 Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 701230
  • Size 100µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Lymphocyte stimulatory factor 1; IGG1 induction factor; B-cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
701230 309.00
Check your price
  • IL-4 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710041
  • Size 100µg
Applications
Western Blot (4-5 ug/ml)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710041 309.00
Check your price
  • IL-4 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIL4I
  • Size 500ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIL4I 309.00
Check your price
  • IL-4 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M2IL4I
  • Size 500ug
Applications
ELISA (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
M2IL4I 299.00
Check your price
  • IL-4 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # PA547930
  • Size 100µg
Applications
Neutralization (2-8 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Lymphocyte stimulatory factor 1; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547930 349.00
Check your price
  • IL-4 Polyclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Goat
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # PA547853
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (1-5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Lymphocyte stimulatory factor 1; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547853 349.00
Check your price
  • IL-4 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Cat
  • Conjugate Unconjugated
  • Catalog # PA547980
  • Size 100µg
Applications
Neutralization (2-8 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Lymphocyte stimulatory factor 1; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547980 349.00
Check your price
  • Phospho-IL4R (Tyr497) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538614
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538614 309.00
Check your price
  • IL4R Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538615
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538615 309.00
Check your price
  • IL-4 Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MM450DB
  • Size 250ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Lymphocyte stimulatory factor 1; IGG1 induction factor; B-cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MM450DB 375.00
Check your price
  • IL-4 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A18491
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18491 117.00
Check your price
  • IL4R Monoclonal Antibody for Western Blot, IHC, Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523814
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (8-25 µg/ml), Neutralization (3-6 ng/ml), Western Blot (1 µg/ml)
Alternate names:
Interleukin-4 receptor subunit alpha, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4R-alpha, IL-4RA, CD124, Soluble interleukin-4 receptor subunit alpha, Soluble IL-4 receptor subunit alpha, Soluble IL-4R-alpha, sIL4Ralpha/prot, IL-4-binding protein, IL4-BP, IL4R, IL4RA, ORF Names: 582J2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523814 275.00
Check your price
  • IL-4 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # A18500
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Lymphocyte stimulatory factor 1; IGG1 induction factor; B-cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18500 172.00
Check your price
  • IL-4 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A18490
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Lymphocyte stimulatory factor 1; IGG1 induction factor; B-cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18490 117.00
Check your price
  • IL-4 Monoclonal Antibody for Western Blot
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MM450D
  • Size 500ug
Applications
Western Blot (Assay Dependent)
Alternate names:
Lymphocyte stimulatory factor 1; IGG1 induction factor; B-cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MM450D 390.00
Check your price
  • IL-4 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA523753
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Lymphocyte stimulatory factor 1; B-cell stimulatory factor 1; B-cell growth factor 1; B-cell IGG differentiation factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523753 349.00
Check your price
  • IL-4 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA523834
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Lymphocyte stimulatory factor 1; B-cell stimulatory factor 1; B-cell growth factor 1; B-cell IGG differentiation factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523834 275.00
Check your price
  • IL-4 Monoclonal Antibody, PE-Cyanine7 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE-Cyanine7
  • Catalog # A18505
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18505 135.00
Check your price
  • IL-4 Monoclonal Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # AHC0749
  • Size 100µg
Applications
ELISA (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHC0749 304.00
Check your price
  • IL-4 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHC0642
  • Size 500µg
Applications
ELISA (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHC0642 304.00
Check your price