1 - 4 of 4 products displayed
  • Importin 9 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA141395
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Ran-binding protein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141395 309.00
Check your price
  • Importin 9 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA141598
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Ran-binding protein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141598 309.00
Check your price
  • Importin 9 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA525477
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ran-binding protein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525477 349.00
Check your price
  • CRM1 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1083
  • Size 100µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (1:500-1:5000)
Alternate names:
Chromosome region maintenance 1 protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1083 375.00
Check your price