1 - 17 of 17 products displayed
  • KPNA4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511810
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511810 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521033
  • Size 100ug
Applications
Immunocytochemistry (2.5 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521033 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132097
  • Size 100µg
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (0.2-1 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132097 349.00
Check your price
  • KPNA3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521035
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
SRP1-gamma; Karyopherin subunit alpha-3; Importin alpha Q2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521035 349.00
Check your price
  • KPNA3 Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132096
  • Size 100ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (3-5 µg/ml), Western Blot (0.1-0.5 µg/ml)
Alternate names:
SRP1-gamma; Karyopherin subunit alpha-3; Importin alpha Q2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132096 375.00
Check your price
  • KPNA2 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA127857
  • Size 50µg
Applications
Western Blot (1:2000)
Alternate names:
SRP1-alpha; RAG cohort protein 1; Pore targeting complex 58 kDa subunit; Pendulin; Karyopherin subunit alpha-2; Importin alpha P1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127857 425.00
Check your price
  • KPNA3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530075
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
SRP1-gamma; Karyopherin subunit alpha-3; Importin alpha Q2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530075 349.00
Check your price
  • KPNA3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528415
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
SRP1-gamma; Karyopherin subunit alpha-3; Importin alpha Q2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528415 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518239
  • Size 100ug
Applications
Immunohistochemistry (3µg/ml), Western Blot (0.2-1 ug/ml)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518239 349.00
Check your price
  • KPNA2 Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Goat
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132094
  • Size 100ug
Applications
ELISA (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (0.1-0.5 µg/ml)
Alternate names:
SRP1-alpha; RAG cohort protein 1; Pore targeting complex 58 kDa subunit; Pendulin; Karyopherin subunit alpha-2; Importin alpha P1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132094 399.00
Check your price
  • KPNA2 Polyclonal Antibody for Western Blot, IF, ICC and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518237
  • Size 100ug
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Western Blot (1-3 µg/ml)
Alternate names:
SRP1-alpha; RAG cohort protein 1; Karyopherin subunit alpha-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518237 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521749
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521749 349.00
Check your price
  • KPNA3 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), IP and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518238
  • Size 100ug
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (5-10µg/ml), Immunoprecipitation (Assay-dependent), Western Blot (0.1-0.5 ug/ml)
Alternate names:
SRP1-gamma; Karyopherin subunit alpha-3; Importin alpha Q2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518238 349.00
Check your price
  • KPNA1 Monoclonal Antibody for Western Blot, IF, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 370800
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
SRP1-beta; RAG cohort protein 2; Nucleoprotein interactor 1; Karyopherin subunit alpha-1; Importin alpha-S1
Catalog # Size Price (AUD) Your Price (AUD) Qty
370800 303.00
Check your price
  • TNPO1 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524581
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
M9 region interaction protein; Karyopherin beta-2; Importin beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524581 349.00
Check your price
  • RAN Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Bovine, Hamster, Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA120581
  • Size 50µL
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (0.5-1 µg/ml)
Alternate names:
Ras-related nuclear protein; GTPase Ran; Ras-like protein TC4; Androgen receptor-associated protein 24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120581 349.00
Check your price
  • RAN Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532570
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Ras-related nuclear protein; Ras-like protein TC4; GTPase Ran; Androgen receptor-associated protein 24
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532570 349.00
Check your price