1 - 2 of 2 products displayed
  • RPS3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517214
  • Size 100µL
Applications
Immunocytochemistry (1:25), Immunofluorescence (1:25), Western Blot (1:1000)
Alternate names:
40S ribosomal protein S3; IMR-90 ribosomal protein S3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517214 275.00
Check your price
  • RPS3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527974
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
S3, 40S ribosomal protein S3, IMR-90 ribosomal protein S3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527974 349.00
Check your price