1 - 30 of 30 products displayed
  • Influenza A H3N2 Monoclonal Antibody for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA183743
  • Size 100 µg
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
Influenza H3N2, H3N2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183743 600.00
Check your price
  • Influenza A H3N2 Polyclonal Antibody for Western Blot, IF, ICC, IHC and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA127282
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Assay-dependent), Western Blot (Assay-dependent)
Alternate names:
Influenza H3N2, H3N2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127282 375.00
Check your price
  • Influenza A H3N2 Polyclonal Antibody for IF
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA17222
  • Size 1 mL
Applications
Immunofluorescence (1:1000)
Alternate names:
Influenza H3N2, H3N2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA17222 375.00
Check your price
  • Influenza A H3N2 Polyclonal Antibody, Biotin conjugate for IF, ICC and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # PA126179
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1000)
Alternate names:
Influenza H3N2, H3N2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126179 399.00
Check your price
  • Influenza A H3N2 HA Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534930
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Influenza H3N2, H3N2 HA, H3N2 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534930 349.00
Check your price
  • Swine Influenza H1N1 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA520714
  • Size 100 ug
Applications
Western Blot (Assay Dependent)
Alternate names:
Influenza H1N1, H1N1 HA, H1N1 hemagglutinin, swine H1N1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520714 349.00
Check your price
  • Avian Influenza H5N1 Neuraminidase Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534949
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
H5N1, Influenza A Neuraminidase, Avian H5N1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534949 349.00
Check your price
  • Avian Influenza H5N3 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534921
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
H5N3, Influenza A Hemagglutinin, Avian H5N3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534921 349.00
Check your price
  • Influenza A PB1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534914
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Influenza PB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534914 349.00
Check your price
  • Influenza A H3N2 M1 Polyclonal Antibody for IF
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA17223
  • Size 1 mL
Applications
Immunofluorescence (1:1000)
Alternate names:
Influenza H3N2, H3N2, H3N2 Matrix protein, H3N2 M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA17223 309.00
Check your price
  • Influenza A H3N2 M1 Polyclonal Antibody, Biotin conjugate for Western Blot, IF, IHC and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # PA173052
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Influenza H3N2, H3N2, H3N2 Matrix protein, H3N2 M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173052 309.00
Check your price
  • Influenza A H3N2 M1 Polyclonal Antibody, FITC conjugate for IF
  • Host Goat
  • Target Species Virus
  • Conjugate FITC
  • Catalog # PA173050
  • Size 1 mL
Applications
Immunofluorescence (1:10)
Alternate names:
Influenza H3N2, H3N2, H3N2 Matrix protein, H3N2 M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173050 349.00
Check your price
  • Influenza A H3N2 M1 Polyclonal Antibody, HRP conjugate for ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA173051
  • Size 1 mL
Applications
ELISA (1:200)
Alternate names:
Influenza H3N2, H3N2, H3N2 Matrix protein, H3N2 M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173051 375.00
Check your price
  • Influenza A H3N2 Virions Polyclonal Antibody, Biotin conjugate for Western Blot, IF, IHC and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # PA173049
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Influenza H3N2, H3N2, H3N2 virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173049 309.00
Check your price
  • Influenza A H3N2 Virions Polyclonal Antibody, FITC conjugate for IF
  • Host Goat
  • Target Species Virus
  • Conjugate FITC
  • Catalog # PA173047
  • Size 1 mL
Applications
Immunofluorescence (1:10-1:50)
Alternate names:
Influenza H3N2, H3N2, H3N2 virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173047 375.00
Check your price
  • Influenza A H3N2 Virions Polyclonal Antibody, HRP conjugate for ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA173048
  • Size 1 mL
Applications
ELISA (1:200)
Alternate names:
Influenza H3N2, H3N2, H3N2 virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173048 309.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF and ICC
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA123054
  • Size 500 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123054 399.00
Check your price
  • Influenza A M1 Polyclonal Antibody, HRP conjugate for ICC and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA127357
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent)
Alternate names:
Influenza Matrix protein M1, Influenza M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127357 309.00
Check your price
  • Influenza A M1 Monoclonal Antibody for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA180736
  • Size 1 mg
Applications
Immunofluorescence (1/100), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Influenza Matrix protein M1, Influenza M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180736 425.00
Check your price
  • Influenza A M1 Polyclonal Antibody for IF
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA185626
  • Size 1 mL
Applications
Immunofluorescence (1:50-1:160)
Alternate names:
Influenza Matrix protein M1, Influenza M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185626 399.00
Check your price
  • Influenza A M1 Polyclonal Antibody for IF, ICC and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA127326
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Influenza Matrix protein M1, Influenza M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127326 375.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA126958
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126958 375.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516291
  • Size 2 mg
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516291 600.00
Check your price
  • Influenza A H1N1 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534920
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Influenza H1N1, H1N1 HA, H1N1 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534920 349.00
Check your price
  • Influenza A H1N1 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534919
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Influenza H1N1, H1N1 HA, H1N1 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534919 349.00
Check your price
  • Influenza A H1N1 HA Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534929
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Influenza H1N1, H1N1 HA, H1N1 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534929 349.00
Check your price
  • Influenza A H9N2 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534925
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Influenza H9N2, H9N2 HA, H9N2 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534925 349.00
Check your price
  • Influenza A H9N2 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534926
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Influenza H9N2, H9N2 HA, H9N2 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534926 349.00
Check your price
  • Influenza A H7N7 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534923
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Influenza H7N7, H7N7 HA, H7N7 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534923 349.00
Check your price
  • Influenza A H9N2 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534924
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Influenza H9N2, H9N2 HA, H9N2 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534924 349.00
Check your price