1 - 11 of 11 products displayed
  • Influenza A M2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA532233
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Influenza Matrix protein M2, Influenza M2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532233 349.00
Check your price
  • Influenza A M1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA532222
  • Size 100µL
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
Influenza Matrix protein M1, Influenza M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532222 349.00
Check your price
  • Influenza A M1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA532253
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Influenza Matrix protein M1, Influenza M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532253 349.00
Check your price
  • Influenza A M2 Monoclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA1082
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1 ug/ml), Immunofluorescence (1 ug/ml), Immunoprecipitation (10 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Influenza Matrix protein M2, Influenza M2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1082 375.00
Check your price
  • Influenza A H9N2 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534925
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Influenza H9N2, H9N2 HA, H9N2 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534925 349.00
Check your price
  • Influenza A H7N7 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534923
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Influenza H7N7, H7N7 HA, H7N7 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534923 349.00
Check your price
  • Influenza A H9N2 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534924
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Influenza H9N2, H9N2 HA, H9N2 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534924 349.00
Check your price
  • Influenza A H7N7 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534922
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Influenza H7N7, H7N7 HA, H7N7 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534922 349.00
Check your price
  • Influenza A H3N2 HA Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534930
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Influenza H3N2, H3N2 HA, H3N2 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534930 349.00
Check your price
  • Influenza A H9N2 HA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA534926
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Influenza H9N2, H9N2 HA, H9N2 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534926 349.00
Check your price
  • Avian Influenza A M2 Synthetic Peptide for BLOCK and Ctrl
  • Host N/A
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PEP0533
  • Size 50ug
Applications
Blocking Assay (Assay Dependent), Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP0533 85.75
Check your price