1 - 30 of 30 products displayed
  • Influenza A H5 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA518212
  • Size 100µg
Applications
ELISA (1:20-1:200)
Alternate names:
Influenza H5, Influenza NP-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518212 287.00
Check your price
  • Influenza A NP Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA532242
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532242 349.00
Check your price
  • Influenza A H5 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA181928
  • Size 200µg
Applications
ELISA (Assay Dependent)
Alternate names:
Influenza H5, Influenza NP-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181928 349.00
Check your price
  • Influenza B NP Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA515375
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515375 349.00
Check your price
  • Influenza A NP Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA517101
  • Size 100µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517101 309.00
Check your price
  • Influenza B NP Monoclonal Antibody for Western Blot, IF, ICC, IHC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1160302
  • Size 200µl
Applications
ELISA (Assay dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Immunohistochemistry (Assay dependent), Western Blot (1:40)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1160302 550.00
Check your price
  • Influenza B NP Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1160202
  • Size 200µl
Applications
ELISA (Assay dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Western Blot (1:20)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1160202 600.00
Check your price
  • Influenza B NP Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1160102
  • Size 200µl
Applications
ELISA (1:200), Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Western Blot (Assay dependent)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1160102 600.00
Check your price
  • Influenza A NP Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1560102
  • Size 200µl
Applications
ELISA (Assay dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Western Blot (Assay dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1560102 600.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF, ICC, IHC (F) and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17057
  • Size 1mL
Applications
ELISA (1:20), Immunocytochemistry (1:20), Immunofluorescence (1:10), Immunohistochemistry (Frozen) (1:10)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17057 349.00
Check your price
  • Influenza A H6N1 NP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA141060
  • Size 100µg
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Influenza H6N1, H6N1 NP, H6N1 Nuclear Protein,
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141060 325.00
Check your price
  • Influenza A H6N1 NP Polyclonal Antibody for IF, ICC, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA141156
  • Size 100µg
Applications
ELISA (0.1-1 µg/ml), Immunocytochemistry (1:10), Immunofluorescence (1:10), Immunohistochemistry (Paraffin) (1:10-1:500)
Alternate names:
Influenza H6N1, H6N1 NP, H6N1 Nuclear Protein,
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141156 309.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF and ICC
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA123054
  • Size 500µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123054 399.00
Check your price
  • Influenza B NP Monoclonal Antibody for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17309
  • Size 100µg
Applications
Immunofluorescence (1:10)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17309 309.00
Check your price
  • Influenza B NP Monoclonal Antibody for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17308
  • Size 100µg
Applications
Immunofluorescence (1:10-1:50)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17308 309.00
Check your price
  • Influenza B NP Monoclonal Antibody for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17310
  • Size 100µg
Applications
Immunofluorescence (1:10)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17310 309.00
Check your price
  • Influenza A NP Monoclonal Antibody, FITC conjugate for IF, IHC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17321
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (1:5), Immunohistochemistry (Assay Dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17321 349.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17323
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17323 375.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17325
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17325 309.00
Check your price
  • Swine Influenza H1N1 NP Polyclonal Antibody for IF
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA520943
  • Size 100ug
Applications
Immunofluorescence (20 ug/ml)
Alternate names:
Influenza H1N1, H1N1 Nuclear Protein, swine H1N1 Nuclear Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520943 349.00
Check your price
  • Influenza B NP Monoclonal Antibody, FITC conjugate for IF and ICC
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17306
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (1:5-1:20)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17306 375.00
Check your price
  • Influenza B NP Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA180712
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180712 375.00
Check your price
  • Influenza A NP Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA141244
  • Size 100µg
Applications
ELISA (0.1-2 µg/ml)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141244 309.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516291
  • Size 2mg
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516291 600.00
Check your price
  • Influenza A NP Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141071
  • Size 100µg
Applications
ELISA (1:100-1:2000), Western Blot (1-3 µg/ml)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141071 375.00
Check your price
  • Influenza B NP Monoclonal Antibody for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17058
  • Size 1mL
Applications
Immunofluorescence (1:10)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17058 309.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF, IHC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17056
  • Size 1mL
Applications
ELISA (1:20 -1:200), Immunofluorescence (1:10 -1:50), Immunohistochemistry (1:10-1:50)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17056 349.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF and IHC (F)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA183740
  • Size 100µg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183740 550.00
Check your price
  • Influenza A NP Monoclonal Antibody, FITC conjugate for IF, Flow and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17322
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17322 375.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17324
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17324 349.00
Check your price