1 - 14 of 14 products displayed
  • INHA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529231
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
A-inhibin subunit, inhibin alpha chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529231 349.00
Check your price
  • INHA Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532452
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
A-inhibin subunit, inhibin alpha chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532452 349.00
Check your price
  • INHA Monoclonal Antibody for IF, ICC and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515315
  • Size 100 ug
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000)
Alternate names:
INHA ;inhibin, alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515315 349.00
Check your price
  • INHA Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513681
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Inhibin alpha chain; INHA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513681 349.00
Check your price
  • INHA Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515703
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
INHA; inhibin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515703 349.00
Check your price
  • INHBA Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521939
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Erythroid differentiation protein; Activin beta-A chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521939 349.00
Check your price
  • INHBB Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528814
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Activin beta-B chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528814 349.00
Check your price
  • INHBC Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA536525
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Activin beta-C chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536525 309.00
Check your price
  • INHBA Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547004
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (2-6 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Activin beta-A chain; Erythroid differentiation protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547004 375.00
Check your price
  • INHBA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Goat, Human, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA544198
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Activin beta-A chain; Erythroid differentiation protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544198 309.00
Check your price
  • INHBC Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA548063
  • Size 100 µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Activin beta-C chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548063 309.00
Check your price
  • Inhibin a (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75632
  • Size 10 µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75632 209.00
Check your price
  • Inhibin Beta A Chain (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75635
  • Size 10 µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75635 225.00
Check your price
  • Inhibin beta A chain (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75636
  • Size 100 µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75636 552.00
Check your price