1 - 7 of 7 products displayed
  • INHA Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532452
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
A-inhibin subunit, inhibin alpha chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532452 349.00
Check your price
  • INHA Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529231
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
A-inhibin subunit, inhibin alpha chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529231 349.00
Check your price
  • Human Activin A Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75636
  • Size 100 µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75636 552.00
Check your price
  • Human INHA Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75632
  • Size 10 µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75632 209.00
Check your price
  • Human Activin A Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75637
  • Size 1 mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75637 2
Check your price
  • Human INHA Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75634
  • Size 1 mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75634 2
Check your price
  • Human INHA Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75633
  • Size 100 µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75633 542.00
Check your price