1 - 30 of 30 products displayed
  • ING1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA519208
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a, growth inhibitor ING1, growth inhibitory protein ING1, inhibitor of growth protein 1, tumor suppressor ING1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA519208 309.00
Check your price
  • ING4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 407700
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
p29ING4
Catalog # Size Price (AUD) Your Price (AUD) Qty
407700 309.00
Check your price
  • ING1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547772
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a, growth inhibitor ING1, growth inhibitory protein ING1, inhibitor of growth protein 1, tumor suppressor ING1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547772 349.00
Check your price
  • ING1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527716
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a, growth inhibitor ING1, growth inhibitory protein ING1, inhibitor of growth protein 1, tumor suppressor ING1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527716 349.00
Check your price
  • Amyloid Precursor Protein Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA125489
  • Size 50µg
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (5-10 µg/ml)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125489 475.00
Check your price
  • ING2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131556
  • Size 50µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Western Blot (2-4 µg/ml)
Alternate names:
p33ING2; p32; Inhibitor of growth 1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131556 349.00
Check your price
  • Amyloid Precursor Protein Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112528
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112528 309.00
Check your price
  • Amyloid Precursor Protein Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112527
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112527 349.00
Check your price
  • GADD34 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132203
  • Size 50µg
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Myeloid differentiation primary response protein MyD116 homolog; Growth arrest and DNA damage-inducible protein GADD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132203 399.00
Check your price
  • Apolipoprotein J Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125833
  • Size 50µg
Applications
Immunocytochemistry (1:2,000), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:2000)
Alternate names:
Testosterone-repressed prostate message 2; NA1/NA2; Ku70-binding protein 1; Complement-associated protein SP-40,40; Complement cytolysis inhibitor; Apolipoprotein J; Aging-associated gene 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125833 399.00
Check your price
  • p19 INK4d Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 378700
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p19-INK4d
Catalog # Size Price (AUD) Your Price (AUD) Qty
378700 304.00
Check your price
  • IGF1 (human) for Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP8606
  • Size 100µg
Applications
Control (Assay Dependent)
Alternate names:
Somatomedin-C; Mechano growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8606 314.00
Check your price
  • IGF1 (mouse) for Ctrl
  • Host E. coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP8603
  • Size 50µg
Applications
Control (Assay Dependent)
Alternate names:
Somatomedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8603 269.00
Check your price
  • IGF1 (rat) for Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP8604
  • Size 50µg
Applications
Control (Assay Dependent)
Alternate names:
Somatomedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8604 301.00
Check your price
  • Muscle-Cardiac FABP (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75547
  • Size 1mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Muscle fatty acid-binding protein; Mammary-derived growth inhibitor; Heart-type fatty acid-binding protein; Fatty acid-binding protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75547 2,500.00
Check your price
  • Human Interleukin 11 (IL-11) for Western Blot, IP, ELISA and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RIL1110
  • Size 10ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Adipogenesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIL1110 309.00
Check your price
  • Amyloid Precursor Protein Monoclonal Antibody for Western Blot, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # OMA103132
  • Size 50µg
Applications
ELISA (10 µg/ml), Immunocytochemistry (0.5-5 µg/ml), Immunoprecipitation (25 µg/ml*), Western Blot (5 µg/ml)
Alternate names:
Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI; ABPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
OMA103132 349.00
Check your price
  • Amyloid Precursor Protein Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532262
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI; ABPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532262 349.00
Check your price
  • Amyloid Precursor Protein Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14648
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14648 375.00
Check your price
  • Amyloid Precursor Protein Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA184165
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1/500-1/1000)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184165 399.00
Check your price
  • Amyloid Precursor Protein Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137099
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI; ABPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137099 475.00
Check your price
  • ING2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518054
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
p33ING2; p32; Inhibitor of growth 1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518054 349.00
Check your price
  • PPP1R1C Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539369
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Inhibitor-5 of protein phosphatase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539369 309.00
Check your price
  • Nogo-A/B Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141220
  • Size 200µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:500-1:2000)
Alternate names:
Reticulon-5; RTN-x; Neuroendocrine-specific protein C homolog; Neuroendocrine-specific protein; Neurite outgrowth inhibitor; Foocen; Glut4 vesicle 20 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141220 309.00
Check your price
  • PPP1R13L Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540602
  • Size 100µl
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:100), Western Blot (1:1000)
Alternate names:
PPP1R13B-like protein; NFkB-interacting protein 1; Inhibitor of ASPP protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540602 309.00
Check your price
  • p19 INK4d Oligoclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710851
  • Size 100µg
Applications
Immunoprecipitation (2 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
p19-INK4d
Catalog # Size Price (AUD) Your Price (AUD) Qty
710851 309.00
Check your price
  • KAZALD1 Polyclonal Antibody for Western Blot
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547701
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Bono1, FKSG28, FKSG40, KAZALD1, bone- and odontoblast-expressed gene 1, BONO1, FKSG40, FLJ24094, IGFBP-related protein 10, IGFBP-rP10, Kazal-type serine peptidase inhibitor domain 1, Kazal-type serine protease inhibitor domain 1, kazal-type serine protease inhibitor domain-containing protein 1, novel kazal-type serine protease inhibitor domain and immunoglobulin domain containing protein, UNQ2945/PRO21184
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547701 349.00
Check your price
  • Apolipoprotein J Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534651
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Sulfated glycoprotein 2; Clustrin; Apolipoprotein J; Testosterone-repressed prostate message 2; NA1/NA2; Ku70-binding protein 1; Complement-associated protein SP-40,40; Complement cytolysis inhibitor; Aging-associated gene 4 protein; Testosterone repressed prostate message 2; Dimeric acid glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534651 349.00
Check your price
  • Apolipoprotein J Monoclonal Antibody for Western Blot, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183879
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Testosterone-repressed prostate message 2; NA1/NA2; Ku70-binding protein 1; Complement-associated protein SP-40,40; Complement cytolysis inhibitor; Apolipoprotein J; Aging-associated gene 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183879 600.00
Check your price
  • Amyloid Precursor Protein Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA133107
  • Size 50µL
Applications
Immunoprecipitation (Neat), Western Blot (1:500-1:1,000)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA133107 449.00
Check your price