1 - 21 of 21 products displayed
  • iNOS Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516422
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516422 499.00
Check your price
  • iNOS Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA3030A
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Frozen) (1:500), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:2,000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3030A 375.00
Check your price
  • iNOS Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121054
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121054 399.00
Check your price
  • iNOS Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA516855
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516855 309.00
Check your price
  • iNOS Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516524
  • Size 500 µL
Applications
Immunohistochemistry (Assay Dependent)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516524 349.00
Check your price
  • iNOS Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1036
  • Size 200 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (1:20), Western Blot (1:200-1:800)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1036 375.00
Check your price
  • Phospho-iNOS (Tyr151) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537667
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537667 309.00
Check your price
  • Phospho-iNOS (Tyr151) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537666
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537666 309.00
Check your price
  • iNOS Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA517106
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517106 275.00
Check your price
  • iNOS Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA137925
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137925 389.00
Check your price
  • iNOS Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517139
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517139 309.00
Check your price
  • iNOS Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137924
  • Size 7 mL
Applications
Immunohistochemistry (Paraffin) (Ready-to-use)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137924 225.00
Check your price
  • iNOS Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA146251
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146251 309.00
Check your price
  • uNOS Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Arthropod, Bovine, Fruit fly, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA1039
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (1:100), Western Blot (1:1,000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1039 375.00
Check your price
  • uNOS Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138835
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138835 399.00
Check your price
  • uNOS Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA3030
  • Size 200 µL
Applications
Immunocytochemistry (1:20), Immunofluorescence (1:20), Immunohistochemistry (Frozen) (1:100), Immunoprecipitation (Assay Dependent), Western Blot (1:50-1:500)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3030 349.00
Check your price
  • Mouse macrophage iNOS (residues 1131-1144) Synthetic Peptide for Ctrl
  • Host N/A
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PEP008
  • Size 50 µg
Applications
Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP008 99.75
Check your price
  • Phospho-nNOS (Ser1417) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1032
  • Size 100 µg
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Western Blot (1:500-1:5000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS1; Neuronal NOS; NOS type I; NC-NOS; Constitutive NOS; BNOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1032 375.00
Check your price
  • nNOS Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1033
  • Size 100 µg
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1:100-1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS1; Neuronal NOS; NOS type I; NC-NOS; Constitutive NOS; BNOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1033 375.00
Check your price
  • eNOS Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137625
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:70)
Alternate names:
NOS type III; Endothelial NOS; EC-NOS; Constitutive NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137625 399.00
Check your price
  • eNOS Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA516887
  • Size 500 µL
Applications
Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:50-1:500)
Alternate names:
NOS type III; Endothelial NOS; EC-NOS; Constitutive NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516887 309.00
Check your price