1 - 30 of 30 products displayed
  • IGF2 Polyclonal Antibody for Western Blot, ELISA, Neu and IA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124773
  • Size 50 µg
Applications
ELISA (0.5-1 µg/ml), Inhibition Assays (0.025-0.075 µg/ml), Neutralization (0.025-0.075 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
T3M-11-derived growth factor; Somatomedin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124773 349.00
Check your price
  • IGF2 (human) for Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP8652
  • Size 50 µg
Applications
Control (Assay Dependent)
Alternate names:
T3M-11-derived growth factor; Somatomedin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8652 269.00
Check your price
  • IGF2 Polyclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA186837
  • Size 50 µg
Applications
ELISA (0.25-1 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
T3M-11-derived growth factor; Somatomedin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186837 475.00
Check your price
  • IGF2 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517096
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
T3M-11-derived growth factor; Somatomedin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517096 309.00
Check your price
  • IGF2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532547
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
T3M-11-derived growth factor; Somatomedin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532547 349.00
Check your price
  • Pds1 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA512252
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100-200)
Alternate names:
T3M-11-derived growth factor; Somatomedin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512252 349.00
Check your price
  • IGF2 Polyclonal Antibody for Western Blot, IHC (F) and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547946
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.5-2.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Multiplication-stimulating polypeptide; IGF-II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547946 349.00
Check your price
  • IGF1R alpha Monoclonal Antibody, Biotin conjugate for IF, ICC, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513799
  • Size 500 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (2 µg/ml)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513799 399.00
Check your price
  • IGF1R alpha Monoclonal Antibody for Western Blot, IF, ICC, IP and IA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA513817
  • Size 500 µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunoprecipitation (2 µg/ml), Inhibition Assays (1.5 µg/mL), Western Blot (1:10-1:200)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513817 349.00
Check your price
  • IGF1R alpha Monoclonal Antibody for IP and IA
  • Host Mouse
  • Target Species Human, Pig, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA513807
  • Size 500 µL
Applications
Immunoprecipitation (2 µg/ml), Inhibition Assays (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513807 349.00
Check your price
  • IGF1R alpha Monoclonal Antibody for IP and IA
  • Host Mouse
  • Target Species Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA513812
  • Size 500 µL
Applications
Immunoprecipitation (Assay Dependent), Inhibition Assays (1.5 µg/mL)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513812 349.00
Check your price
  • IGF1R alpha Monoclonal Antibody for IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHR0321
  • Size 500 µl
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunoprecipitation (2µg per mg lysate)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHR0321 304.00
Check your price
  • IGF2R Monoclonal Antibody, Alexa Fluor® 647 conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Alexa Fluor® 647
  • Catalog # MA518132
  • Size 100 tests
Applications
Flow Cytometry (4ulx100ul whole blood or 10^6 cells)
Alternate names:
M6P/IGF2 receptor; Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518132 249.00
Check your price
  • IGF2R Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate FITC
  • Catalog # MA110147
  • Size 100 Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
M6P/IGF2 receptor; Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110147 249.00
Check your price
  • IGF2R Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate APC
  • Catalog # MA110146
  • Size 100 Tests
Applications
Flow Cytometry (10 ul/10^6 cells)
Alternate names:
M6P/IGF2 receptor; Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110146 309.00
Check your price
  • IGF2R Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate PE
  • Catalog # MA110148
  • Size 100 Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
M6P/IGF2 receptor; Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110148 275.00
Check your price
  • IGF2R Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA119252
  • Size 100 µg
Applications
Flow Cytometry (1-2µg/ml), Western Blot (Assay Dependent)
Alternate names:
M6P/IGF2 receptor; Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119252 199.00
Check your price
  • IGFBP3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518791
  • Size 100 ug
Applications
Western Blot (0.02-0.06 ug/ml)
Alternate names:
IGFBP3; insulin-like growth factor binding protein 3; tcag7.703; BP-53; IBP3; IGF-binding protein 3; acid stable subunit of the 140 K IGF complex ; binding protein 29; binding protein 53; growth hormone-dependent binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518791 349.00
Check your price
  • Phospho-IGFBP3 (Ser183) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537606
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
BP-53, IBP3, IBP-3, IGF-binding protein 3, IGFBP-3, acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, insulin-like growth factor-binding protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537606 309.00
Check your price
  • IGFBP3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527190
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
BP-53, IBP3, IBP-3, IGF-binding protein 3, IGFBP-3, acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, insulin-like growth factor-binding protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527190 349.00
Check your price
  • IGFBP3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529711
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
BP-53, IBP3, IBP-3, IGF-binding protein 3, IGFBP-3, acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, insulin-like growth factor-binding protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529711 349.00
Check your price
  • IGFBP5 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523872
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
IBP 5, IBP-5, IBP5, IBP5, IGF binding protein 5, IGF BP5, IGF-binding protein 5, IGFBP 5, IGFBP-5, IGFBP5, Insulin Like Growth Factor Binding Protein 5, Insulin-like growth factor-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523872 275.00
Check your price
  • IGFBP5 Polyclonal Antibody for Western Blot, IHC (F) and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547775
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IBP 5, IBP-5, IBP5, IBP5, IGF binding protein 5, IGF BP5, IGF-binding protein 5, IGFBP 5, IGFBP-5, IGFBP5, Insulin Like Growth Factor Binding Protein 5, Insulin-like growth factor-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547775 349.00
Check your price
  • IGFBP5 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537369
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
IBP 5, IBP-5, IBP5, IBP5, IGF binding protein 5, IGF BP5, IGF-binding protein 5, IGFBP 5, IGFBP-5, IGFBP5, Insulin Like Growth Factor Binding Protein 5, Insulin-like growth factor-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537369 399.00
Check your price
  • IGFBP4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518928
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
IGFBP4 ; insulin-like growth factor binding protein 4 ; BP-4; HT29-IGFBP; IBP4; IGFBP-4; IGF-binding protein 4; insulin-like growth factor-binding protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518928 349.00
Check your price
  • Phospho-IGF2R (Ser2409) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA537605
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
M6P/IGF2 receptor; Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537605 309.00
Check your price
  • IGF2R Monoclonal Antibody for Western Blot, IF, ICC, IHC (PFA), Flow and IP
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1066
  • Size 100 µg
Applications
Flow Cytometry (1µg per 10^6 cells), Immunocytochemistry (2-5 µg/ml), Immunofluorescence (2-5 µg/ml), Immunohistochemistry (PFA fixed) (Assay Dependent), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor; M6P/IGF2 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1066 375.00
Check your price
  • IGF2R Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # A15744
  • Size 100 tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
M6P/IGF2 receptor; Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15744 139.00
Check your price
  • IGF2R Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # A15710
  • Size 100 tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
M6P/IGF2 receptor; Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15710 169.00
Check your price
  • IGF2R Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A15775
  • Size 100 tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
M6P/IGF2 receptor; Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15775 169.00
Check your price