1 - 30 of 30 products displayed
  • INSR beta Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP, ELISA and AP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA513778
  • Size 500µL
Applications
Affinity Purification (Assay Dependent), ELISA (Assay Dependent), Flow Cytometry (2 ug/test), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunoprecipitation (2 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
CD220, CD220 beta, HHF5, IR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513778 349.00
Check your price
  • INSR beta Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # AHR0271
  • Size 500µl
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
CD220, CD220 beta, HHF5, IR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHR0271 304.00
Check your price
  • Phospho-INSR (Tyr1328) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44807G
  • Size 10blots
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (Assay Dependent)
Alternate names:
CD220, CD220 beta, HHF5, IR beta, Insulin Receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
44807G 309.00
Check your price
  • Phospho-INSR (Tyr1334) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44809G
  • Size 10blots
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (Assay Dependent)
Alternate names:
CD220, CD220 beta, HHF5, IR beta, Insulin Receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
44809G 304.00
Check your price
  • Phospho-INSR (Tyr972) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44800G
  • Size 10blots
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (Assay Dependent)
Alternate names:
CD220, CD220 beta, HHF5, IR beta, Insulin Receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
44800G 304.00
Check your price
  • INSR beta Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710076
  • Size 100µg
Applications
Western Blot (2-5 ug/ml)
Alternate names:
CD220, HHF5, insulin receptor, INSR, CD221, IGF-I receptor, insulin-like growth factor I receptor, soluble IGF1R variant 1, soluble IGF1R variant 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710076 309.00
Check your price
  • PKC beta Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529680
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
PKC-beta, PKCB, PRKCB1, PRKCB2, PKC-B, protein kinase C beta type, protein kinase C, beta 1 polypeptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529680 349.00
Check your price
  • Phospho-PKC beta (Thr641) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537692
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Protein kinase C beta type, PKC-B, PKC-beta, PRKCB, PKCB, PRKCB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537692 309.00
Check your price
  • Phospho-IR/IGF1R (Tyr1135, Tyr1136, Tyr1150, Tyr1151) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515148
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515148 309.00
Check your price
  • beta Catenin Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA516429
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (10 µg/ml), Western Blot (5-10 µg/mL)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516429 349.00
Check your price
  • Phospho-beta Catenin (Ser33, Ser37) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 486000
  • Size 100µL
Applications
Western Blot (1:1,000)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
486000 309.00
Check your price
  • beta Catenin Polyclonal Antibody for Western Blot, IF, IHC (F), IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rodent, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA519469
  • Size 100µg
Applications
ELISA (0.1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (0.25 µg/ml)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519469 349.00
Check your price
  • Phospho-PKC beta-2 (Thr641) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA122053
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122053 381.00
Check your price
  • IL-1 beta Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M421B
  • Size 500ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
M421B 349.00
Check your price
  • beta Catenin Polyclonal Antibody for IF, ICC, IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 712700
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:20-1:200), Immunoprecipitation (Assay Dependent)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
712700 305.00
Check your price
  • IL-1 beta Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA523657
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523657 349.00
Check your price
  • IL-1 beta Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523546
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523546 239.00
Check your price
  • IL-1 beta Monoclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523691
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (0.05-0.2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523691 375.00
Check your price
  • IL-1 beta Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # A18465
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18465 106.00
Check your price
  • IL-1 beta Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A18489
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18489 106.00
Check your price
  • IL-1 beta Monoclonal Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M420BB
  • Size 250µg
Applications
ELISA (Assay Dependent)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
M420BB 349.00
Check your price
  • IL-1 beta Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523597
  • Size 100Tests
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523597 289.00
Check your price
  • IL-1 beta Polyclonal Antibody for Western Blot, IHC, ELISA and Neu
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # P420B
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Neutralization (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
P420B 349.00
Check your price
  • IL-1 beta Monoclonal Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # AHC0519
  • Size 100µg
Applications
ELISA (Assay Dependent)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHC0519 305.00
Check your price
  • Phospho-PKA 2 beta pSer114 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat, Bovine, Human, Mouse, Rabbit
  • Conjugate
  • Catalog # PA126735
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126735 Not Orderable Online
Check your price
  • Phospho-INSR (Tyr1158) Oligoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710101
  • Size 100µg
Applications
Western Blot (1-5 ug/ml)
Alternate names:
CD220, HHF5, insulin receptor, INSR, CD221, IGF-I receptor, insulin-like growth factor I receptor, soluble IGF1R variant 1, soluble IGF1R variant 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710101 309.00
Check your price
  • Phospho-INSR (Tyr1158) Monoclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701070
  • Size 100µg
Applications
Flow Cytometry (3-5 µg/million cells), Western Blot (1-3 ug/ml)
Alternate names:
CD220, HHF5, insulin receptor, INSR, CD221, IGF-I receptor, insulin-like growth factor I receptor, soluble IGF1R variant 1, soluble IGF1R variant 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
701070 309.00
Check your price
  • INSR Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701049
  • Size 100µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
CD220, HHF5, insulin receptor, INSR, CD221, IGF-I receptor, insulin-like growth factor I receptor, soluble IGF1R variant 1, soluble IGF1R variant 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
701049 303.00
Check your price
  • Phospho-IGF1R beta (Tyr1158, Tyr1162, Tyr1163) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA126725
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126725 349.00
Check your price
  • INSR alpha Monoclonal Antibody for Western Blot, IF, ICC, IP, ELISA, FN and IA
  • Host Mouse
  • Target Species Bovine, Human, Sheep, Pig
  • Conjugate Unconjugated
  • Catalog # AHR0221
  • Size 500µl
Applications
ELISA (Assay Dependent), Functional Assay (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunoprecipitation (2µg per mg lysate), Inhibition Assays (Assay Dependent), Western Blot (Assay-Dependent)
Alternate names:
CD220, CD220 beta, HHF5, IR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHR0221 309.00
Check your price