1 - 8 of 8 products displayed
  • IDE Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524064
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Insulysin; Insulin protease; Abeta-degrading protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524064 275.00
Check your price
  • IDE Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546925
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Insulysin; Insulin protease; Abeta-degrading protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546925 375.00
Check your price
  • IDE Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511520
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Insulysin; Insulin protease; Abeta-degrading protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511520 349.00
Check your price
  • IDE Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518753
  • Size 100ug
Applications
Immunohistochemistry (3-6µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Insulysin; Insulin protease; Abeta-degrading protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518753 349.00
Check your price
  • IDE Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA529350
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Insulysin; Insulin protease; Abeta-degrading protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529350 349.00
Check your price
  • IDE Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528191
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Insulysin; Insulin protease; Abeta-degrading protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528191 349.00
Check your price
  • IDE Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529349
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Insulysin; Insulin protease; Abeta-degrading protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529349 349.00
Check your price
  • IDE Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518754
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Insulysin; Insulin protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518754 349.00
Check your price