1 - 30 of 30 products displayed
  • TMP21 Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA146004
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (2 µg/ml)
Alternate names:
p24 family protein delta-1; Transmembrane protein Tmp21; 21 kDa transmembrane-trafficking protein; p24delta; p23; Tmp-21-I; S31III125; Integral membrane protein p23
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146004 349.00
Check your price
  • TMP21 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rabbit, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA146005
  • Size 100 µL
Applications
Immunohistochemistry (1 µg/ml), Western Blot (2 µg/ml)
Alternate names:
p24 family protein delta-1; Transmembrane protein Tmp21; 21 kDa transmembrane-trafficking protein; p24delta; p23; Tmp-21-I; S31III125; Integral membrane protein p23
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146005 309.00
Check your price
  • LIMP2 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA316802
  • Size 100 µL
Applications
Immunocytochemistry (1:50-1:250), Immunofluorescence (1:50-1:250), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Scavenger receptor class B member 2; Lysosome membrane protein II; 85 kDa lysosomal membrane sialoglycoprotein; CD36 antigen-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316802 375.00
Check your price
  • LITAF Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517242
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
p53-induced gene 7 protein; Small integral membrane protein of lysosome/late endosome
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517242 275.00
Check your price
  • Stomatin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530019
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Stomatin; Protein 7.2b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530019 349.00
Check your price
  • Claudin 2 Polyclonal Antibody for Western Blot, IF and ELISA
  • Host Rabbit
  • Target Species Dog, Human, Rat
  • Conjugate Unconjugated
  • Catalog # 516100
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunofluorescence (1-3 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
SP82
Catalog # Size Price (AUD) Your Price (AUD) Qty
516100 303.00
Check your price
  • CUX1 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530003
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
Homeobox protein cux-1; CCAAT displacement protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530003 349.00
Check your price
  • GPR155 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523074
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Western Blot (1:100-1:2000)
Alternate names:
G-protein coupled receptor PGR22
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523074 349.00
Check your price
  • GPR155 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523075
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Western Blot (1:100-1:2000)
Alternate names:
G-protein coupled receptor PGR22
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523075 349.00
Check your price
  • Trans-Golgi Network 38 Polyclonal Antibody for Western Blot, IF, IHC (F) and IM
  • Host Sheep
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA184496
  • Size 100 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1/100-1/200), Immunomicroscopy (Assay Dependent), Western Blot (1/1000)
Alternate names:
TGN38B; TGN38A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184496 550.00
Check your price
  • Caveolin 1 Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Dog, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1064
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunoprecipitation (5 µg), Western Blot (1-2 µg/ml)
Alternate names:
Vesicular integral-membrane protein VIP21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1064 375.00
Check your price
  • SURF4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533063
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
ERV29, surface 4 integral membrane protein, surfeit locus protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533063 349.00
Check your price
  • VIP36 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530418
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Vesicular integral-membrane protein 36; Lectin mannose-binding 2; Glycoprotein GP36b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530418 349.00
Check your price
  • Mitochondria Membrane Integrity Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 457799
  • Size 1 Each
Applications
Western Blot (10.2 ug/ml)
Alternate names:
Ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit; Cytochrome b-c1 complex subunit 4; Complex III subunit IV; Complex III subunit 4; Rotamase F; Cyclophilin F; Cyclophilin D; Outer mitochondrial membrane protein porin 1; Plasmalemmal porin; Brain-derived voltage-dependent anion channel 1; Porin 31HM; Porin 31HL; Mitochondrial cyclophilin; Voltage-dependent anion-selective channel protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
457799 570.00
Check your price
  • NKG2A/NKG2C/NKG2E Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # A15416
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD159A, NKG2, NKG2A, C-lectin type II protein, CD159 antigen-like family member A, NK cell receptor A, NKG2-1/B activating NK receptor, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, natural killer cell l, KLRC1, cd159e, cd159c
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15416 297.00
Check your price
  • NKG2A/NKG2C/NKG2E Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A15417
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD159A, NKG2, NKG2A, C-lectin type II protein, CD159 antigen-like family member A, NK cell receptor A, NKG2-1/B activating NK receptor, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, natural killer cell l, KLRC1
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15417 191.00
Check your price
  • SDHC Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA545466
  • Size 100 µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Succinate-ubiquinone oxidoreductase cytochrome B large subunit; Succinate dehydrogenase complex subunit C; QPs-1; Integral membrane protein CII-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545466 309.00
Check your price
  • PEX11A Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526025
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Protein PEX11 homolog alpha; Peroxisomal biogenesis factor 11A; Peroxin-11A; 28 kDa peroxisomal integral membrane protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526025 349.00
Check your price
  • SLC7A5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533053
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (20 µg/ml)
Alternate names:
y+ system cationic amino acid transporter; Solute carrier family 7 member 5; L-type amino acid transporter 1; Integral membrane protein E16; CD98 light chain; 4F2 light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533053 349.00
Check your price
  • SLC7A5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533052
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (13-26 µg/ml)
Alternate names:
y+ system cationic amino acid transporter; Solute carrier family 7 member 5; L-type amino acid transporter 1; Integral membrane protein E16; CD98 light chain; 4F2 light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533052 349.00
Check your price
  • SLC7A5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534216
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (15 µg/ml)
Alternate names:
y+ system cationic amino acid transporter; Solute carrier family 7 member 5; L-type amino acid transporter 1; Integral membrane protein E16; CD98 light chain; 4F2 light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534216 349.00
Check your price
  • SLC7A5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534215
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (7 µg/ml)
Alternate names:
y+ system cationic amino acid transporter; Solute carrier family 7 member 5; L-type amino acid transporter 1; Integral membrane protein E16; CD98 light chain; 4F2 light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534215 349.00
Check your price
  • SLC7A5 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519101
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
y+ system cationic amino acid transporter; Solute carrier family 7 member 5; L-type amino acid transporter 1; Integral membrane protein E16; CD98 light chain; 4F2 light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519101 349.00
Check your price
  • LIMP2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520544
  • Size 100 ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Scavenger receptor class B member 2; Lysosome membrane protein II; 85 kDa lysosomal membrane sialoglycoprotein; CD36 antigen-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520544 349.00
Check your price
  • LIMP2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520540
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Scavenger receptor class B member 2; Lysosome membrane protein II; 85 kDa lysosomal membrane sialoglycoprotein; CD36 antigen-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520540 349.00
Check your price
  • GPR177 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542570
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.5-1 µg/ml)
Alternate names:
Protein evenness interrupted homolog; Integral membrane protein GPR177; Putative NF-kappa-B-activating protein 373
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542570 309.00
Check your price
  • TPRA1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132706
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Transmembrane protein 227; Integral membrane protein GPR175
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132706 375.00
Check your price
  • TPRA1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534274
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (9 µg/ml)
Alternate names:
Transmembrane protein 227; Integral membrane protein GPR175
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534274 349.00
Check your price
  • TPRA1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534276
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (20 µg/ml)
Alternate names:
Transmembrane protein 227; Integral membrane protein GPR175
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534276 349.00
Check your price
  • TPRA1 Polyclonal Antibody for ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534275
  • Size 50 ug
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (8-19 µg/ml)
Alternate names:
Transmembrane protein 227; Integral membrane protein GPR175
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534275 349.00
Check your price