1 - 30 of 30 products displayed
  • ITGA2 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # MA523532
  • Size 100Tests
Applications
Flow Cytometry (10 µg/10^6 cells)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523532 199.00
Check your price
  • ITGA2 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA523537
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523537 219.00
Check your price
  • ITGA2 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547193
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547193 349.00
Check your price
  • Filamin A alpha-2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA529598
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Non-muscle filamin; Filamin-1; Endothelial actin-binding protein; Alpha-filamin; Actin-binding protein 280
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529598 349.00
Check your price
  • CD103 Polyclonal Antibody, Alexa Fluor® 488 conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # PA546876
  • Size 100Tests
Applications
Flow Cytometry (5 µl/10^6 cells)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546876 199.00
Check your price
  • CD103 Polyclonal Antibody, PE conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # PA546877
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546877 199.00
Check your price
  • CD103 Polyclonal Antibody, APC conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # PA546874
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546874 169.00
Check your price
  • CD103 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547236
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547236 349.00
Check your price
  • alpha Actinin 2/3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538605
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
F-actin cross-linking protein; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin skeletal muscle isoform 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538605 309.00
Check your price
  • ITGA3 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA127601
  • Size 50µg
Applications
Western Blot (1:2000)
Alternate names:
VLA-3 subunit alpha; Galactoprotein B3; FRP-2; CD49 antigen-like family member C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127601 475.00
Check your price
  • ITGA2 Monoclonal Antibody for Western Blot, IF, IHC (F), IHC (P), Flow, IP, ELISA and FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180784
  • Size 200µg
Applications
ELISA (10 ug/ml), Flow Cytometry (30 ug/ml), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (30-40 ug/ml), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (25-50 ug/ml), Western Blot (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180784 399.00
Check your price
  • ITGA4 Monoclonal Antibody, Biotin conjugate for IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517804
  • Size 100ug
Applications
Flow Cytometry (1.0 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517804 119.00
Check your price
  • ITGA4 Monoclonal Antibody, Biotin conjugate for IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517805
  • Size 300ug
Applications
Flow Cytometry (1.0 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517805 275.00
Check your price
  • ITGA4 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517806
  • Size 100ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517806 139.00
Check your price
  • ITGA4 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517807
  • Size 300ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517807 275.00
Check your price
  • ITGA4 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517808
  • Size 50ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517808 119.00
Check your price
  • ITGA4 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517809
  • Size 300ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517809 375.00
Check your price
  • ITGA4 Monoclonal Antibody for IHC (F), Flow and IP
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA170075
  • Size 250ug
Applications
Flow Cytometry (1 ug/1 million cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170075 275.00
Check your price
  • CD11a Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517950
  • Size 300ug
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD11 antigen-like family member A, LFA-1 alpha, LFA-1A, antigen CD11A p180, lymphocyte function-associated antigen 1 alpha polypeptide, integrin alpha-L
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517950 309.00
Check your price
  • CD11a Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517951
  • Size 100ug
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD11 antigen-like family member A, LFA-1 alpha, LFA-1A, antigen CD11A p180, lymphocyte function-associated antigen 1 alpha polypeptide, integrin alpha-L
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517951 139.00
Check your price
  • CD11a Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517952
  • Size 300ug
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD11 antigen-like family member A, LFA-1 alpha, LFA-1A, antigen CD11A p180, lymphocyte function-associated antigen 1 alpha polypeptide, integrin alpha-L
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517952 309.00
Check your price
  • CD11a Monoclonal Antibody for IHC (F), Flow and IP
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA517949
  • Size 250ug
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
CD11 antigen-like family member A, LFA-1 alpha, LFA-1A, antigen CD11A p180, lymphocyte function-associated antigen 1 alpha polypeptide, integrin alpha-L
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517949 225.00
Check your price
  • ITGA4 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # A16315
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16315 93.75
Check your price
  • ITGA4 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A16329
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16329 119.00
Check your price
  • ITGA2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532880
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532880 349.00
Check your price
  • ITGA2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532879
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532879 349.00
Check your price
  • ITGA4 Monoclonal Antibody for IHC, Flow and Neu
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA49D7
  • Size 500ug
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Assay Dependent), Neutralization (Assay Dependent)
Alternate names:
VLA-4 subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA49D7 349.00
Check your price
  • ITGA4 Monoclonal Antibody for IHC (F), Flow, IP and FN
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA182769
  • Size 500µg
Applications
Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182769 550.00
Check your price
  • ITGA2 Monoclonal Antibody for Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA126714
  • Size 50µg
Applications
ELISA (10 µg/ml), Flow Cytometry (Assay Dependent), Immunoprecipitation (25-50 µg/ml)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126714 309.00
Check your price
  • ITGA2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526061
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526061 349.00
Check your price