1 - 30 of 30 products displayed
  • ITGA2 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # MA523532
  • Size 100Tests
Applications
Flow Cytometry (10 µg/10^6 cells)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523532 199.00
Check your price
  • ITGA2 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA523537
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523537 219.00
Check your price
  • ITGA2 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547193
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547193 349.00
Check your price
  • ITGA2 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate PE
  • Catalog # MA516572
  • Size 100TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516572 349.00
Check your price
  • ITGA2 Monoclonal Antibody for IHC (F), Flow and FN
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA516571
  • Size 200ug
Applications
Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (1:500-1:1000)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516571 375.00
Check your price
  • CD103 Polyclonal Antibody, Alexa Fluor® 488 conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # PA546876
  • Size 100Tests
Applications
Flow Cytometry (5 µl/10^6 cells)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546876 199.00
Check your price
  • CD103 Polyclonal Antibody, PE conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # PA546877
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546877 199.00
Check your price
  • CD103 Polyclonal Antibody, APC conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # PA546874
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546874 169.00
Check your price
  • CD103 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547236
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547236 349.00
Check your price
  • ITGA3 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA127601
  • Size 50µg
Applications
Western Blot (1:2000)
Alternate names:
VLA-3 subunit alpha; Galactoprotein B3; FRP-2; CD49 antigen-like family member C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127601 475.00
Check your price
  • ITGA2 Monoclonal Antibody for Western Blot, IF, IHC (F), IHC (P), Flow, IP, ELISA and FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180784
  • Size 200µg
Applications
ELISA (10 ug/ml), Flow Cytometry (30 ug/ml), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (30-40 ug/ml), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (25-50 ug/ml), Western Blot (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180784 399.00
Check your price
  • ITGA2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532880
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532880 349.00
Check your price
  • ITGA2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532879
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532879 349.00
Check your price
  • ITGA2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526061
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526061 349.00
Check your price
  • ITGA2 Monoclonal Antibody for Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA126714
  • Size 50µg
Applications
ELISA (10 µg/ml), Flow Cytometry (Assay Dependent), Immunoprecipitation (25-50 µg/ml)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126714 309.00
Check your price
  • ITGA2 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA180244
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180244 325.00
Check your price
  • ITGA2 Monoclonal Antibody for IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA146035
  • Size 100µg
Applications
Flow Cytometry (1:10-1:1000), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (1:10-1:500)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146035 309.00
Check your price
  • ITGA2 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # A15431
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15431 214.00
Check your price
  • ITGA2 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A15432
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15432 239.00
Check your price
  • ITGA9/ITGB1 Monoclonal Antibody for Flow and IP
  • Host Mouse
  • Target Species Guinea pig, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA180920
  • Size 200µg
Applications
Flow Cytometry (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
VLA-4 subunit beta; Fibronectin receptor subunit beta; Glycoprotein IIa; Integrin alpha-RLC
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180920 550.00
Check your price
  • ITGA2 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # A14740
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14740 79.50
Check your price
  • ITGA2 Monoclonal Antibody, APC-Cyanine7 conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC-Cyanine7
  • Catalog # A15420
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15420 269.00
Check your price
  • ITGA2 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A15419
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15419 239.00
Check your price
  • ITGA2 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # A14742
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14742 99.25
Check your price
  • ITGA2 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A14741
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14741 99.25
Check your price
  • ITGA2 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA516648
  • Size 100ug
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516648 199.00
Check your price
  • ITGA4 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520600
  • Size 100ug
Applications
Immunocytochemistry (2 ug/ml), Immunofluorescence (2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520600 349.00
Check your price
  • ITGA4 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520599
  • Size 100ug
Applications
Immunocytochemistry (2 ug/ml), Immunofluorescence (2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520599 349.00
Check your price
  • ITGA4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520595
  • Size 100ug
Applications
Western Blot (1 µg/ml)
Alternate names:
VLA-4 subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; Integrin alpha-IV; CD49 antigen-like family member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520595 349.00
Check your price
  • Filamin A alpha-2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA529598
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Non-muscle filamin; Filamin-1; Endothelial actin-binding protein; Alpha-filamin; Actin-binding protein 280
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529598 349.00
Check your price