1 - 30 of 30 products displayed
  • IL-1 alpha Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA546955
  • Size 100µg
Applications
Neutralization (0.003-0.009 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL-1 alpha, hematopoietin-1, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha, Hematopoietin-1, IL1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546955 375.00
Check your price
  • IL-1 alpha Monoclonal Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M420AB
  • Size 250ug
Applications
ELISA (Assay Dependent)
Alternate names:
Hematopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
M420AB 349.00
Check your price
  • IL-1 alpha Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M421A
  • Size 500ug
Applications
ELISA (Assay Dependent)
Alternate names:
Hematopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
M421A 349.00
Check your price
  • IL-1 alpha Monoclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523694
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (0.05-0.15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Hematopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523694 349.00
Check your price
  • IL-1 alpha Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710043
  • Size 100µg
Applications
Western Blot (4-5 ug/ml)
Alternate names:
Hematopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710043 309.00
Check your price
  • IL-1 alpha Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M420A
  • Size 500ug
Applications
ELISA (Assay Dependent)
Alternate names:
Hematopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
M420A 275.00
Check your price
  • IL-1 alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # P420A
  • Size 1mg
Applications
Western Blot (Assay Dependent)
Alternate names:
Hematopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
P420A 309.00
Check your price
  • IL-1 alpha Monoclonal Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # AHC0419
  • Size 100µg
Applications
ELISA (Assay Dependent)
Alternate names:
Hematopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHC0419 304.00
Check your price
  • IL-1 alpha Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHC0912
  • Size 500µg
Applications
ELISA (Assay Dependent)
Alternate names:
Hematopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHC0912 309.00
Check your price
  • Human Interleukin 1 alpha (IL-1a) for Western Blot, IP, ELISA and Ctrl
  • Host Yeast
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RIL1AI
  • Size 20ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Hematopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIL1AI 309.00
Check your price
  • IL-1 alpha Monoclonal Antibody, FITC conjugate for Flow
  • Host Hamster
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # A18463
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
IL-1 alpha, hematopoietin-1, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha, Hematopoietin-1, IL1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18463 Not Orderable Online
Check your price
  • IL-1 alpha Monoclonal Antibody for ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523736
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.2-0.6 µg/ml)
Alternate names:
IL-1 alpha, hematopoietin-1, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha, Hematopoietin-1, IL1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523736 349.00
Check your price
  • IL-1 alpha Polyclonal Antibody for Western Blot, IHC (F) and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547011
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.002-0.004 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL-1 alpha, hematopoietin-1, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha, Hematopoietin-1, IL1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547011 375.00
Check your price
  • IL-1 alpha Monoclonal Antibody for ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523828
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.2-0.6 µg/ml)
Alternate names:
IL-1 alpha, hematopoietin-1, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha, Hematopoietin-1, IL1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523828 275.00
Check your price
  • HIF1A Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow and ChIP
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA116601
  • Size 100µL
Applications
ChIP assay (Assay-Dependent), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (5-10 µg/ml), Immunohistochemistry (Paraffin) (5-10 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116601 375.00
Check your price
  • HIF1A Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), IP, GS and IE
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Primate, Pig
  • Conjugate Unconjugated
  • Catalog # MA1516
  • Size 100µL
Applications
Gel Shift (Assay dependent), Immunocytochemistry (1:20-1:200), Immunoelectrophoresis (Assay Dependent), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1516 375.00
Check your price
  • IL-1 alpha Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # PA547876
  • Size 100µg
Applications
Neutralization (0.006-0.012 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL-1 alpha, hematopoietin-1, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha, Hematopoietin-1, IL1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547876 349.00
Check your price
  • HIF1A Polyclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1184
  • Size 100µg
Applications
ELISA (1:1000), Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunoprecipitation (2 µg), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1184 349.00
Check your price
  • HIF1A Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116518
  • Size 100µL
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunohistochemistry (Frozen) (1:100-1:5000), Immunohistochemistry (Paraffin) (1:100-1:5000), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116518 375.00
Check your price
  • HIF1A Monoclonal Antibody, Biotin conjugate for Western Blot, IHC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA516016
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (1:100-1:5000), Immunohistochemistry (Paraffin) (1:100-1:5000), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516016 375.00
Check your price
  • HIF1A Polyclonal Antibody, DyLight 650 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Primate, Rabbit, Rat
  • Conjugate DyLight 650
  • Catalog # PA522777
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522777 375.00
Check your price
  • HIF1A Polyclonal Antibody, DyLight 488 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Primate, Rabbit, Rat
  • Conjugate DyLight 488
  • Catalog # PA522778
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522778 375.00
Check your price
  • HIF1A Monoclonal Antibody, DyLight 550 conjugate for IF, ICC, IHC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate DyLight 550
  • Catalog # MA516020
  • Size 100µL
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunohistochemistry (Frozen) (1:100-1:5000), Immunohistochemistry (Paraffin) (1:100-1:5000), Immunoprecipitation (1:10-1:500)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516020 375.00
Check your price
  • IL-1 alpha Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525921
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Hematopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525921 349.00
Check your price
  • IL-1 alpha Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA175178
  • Size 500µg
Applications
ELISA (Assay Dependent)
Alternate names:
Hematopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175178 1,500.00
Check your price
  • CCL3 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHC6034
  • Size 500µg
Applications
ELISA (Assay Dependent)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHC6034 303.00
Check your price
  • CCL3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129045
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129045 375.00
Check your price
  • CCL3 Polyclonal Antibody for Western Blot, IHC (P), ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547000
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (2-8 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547000 375.00
Check your price
  • HIF1A Polyclonal Antibody, DyLight 550 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Rabbit, Rat, Goat, Hamster, Human, Mouse, Primate
  • Conjugate DyLight 550
  • Catalog # PA522780
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522780 375.00
Check your price
  • HIF1A Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Primate
  • Conjugate Unconjugated
  • Catalog # MA116517
  • Size 100µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1; ARNT-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116517 349.00
Check your price