1 - 27 of 27 products displayed
  • IL1RAPL2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539608
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Three immunoglobulin domain-containing IL-1 receptor-related 1; Interleukin-1 receptor 9; IL1RAPL-2-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539608 309.00
Check your price
  • IL-1RAcP Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528051
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Interleukin-1 receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528051 349.00
Check your price
  • IL1RAPL2 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547039
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Three immunoglobulin domain-containing IL-1 receptor-related 1; Interleukin-1 receptor 9; IL1RAPL-2-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547039 349.00
Check your price
  • IL-1RAcP Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129969
  • Size 50µg
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
Interleukin-1 receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129969 399.00
Check your price
  • IL-1RAcP Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523667
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Interleukin-1 receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523667 349.00
Check your price
  • IL-1RAcP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130896
  • Size 100µg
Applications
Western Blot (Assay dependent)
Alternate names:
Interleukin-1 receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130896 449.00
Check your price
  • IL-1RAcP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519920
  • Size 100ug
Applications
Western Blot (1:1000)
Alternate names:
Interleukin-1 receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519920 349.00
Check your price
  • IL-1RAcP Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519921
  • Size 100ug
Applications
Immunocytochemistry (5 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
Interleukin-1 receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519921 349.00
Check your price
  • IL18RAP Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547087
  • Size 100µg
Applications
Neutralization (0.3-1.2 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-18 receptor beta; Interleukin-18 receptor accessory protein-like; Interleukin-1 receptor 7; IL-1R accessory protein-like; CDw218b; CD218 antigen-like family member B; Accessory protein-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547087 349.00
Check your price
  • IL1R1 Monoclonal Antibody for IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110859
  • Size 200ug
Applications
ELISA (5 µg/m), Immunohistochemistry (10-100 µg/ml)
Alternate names:
p80; Interleukin-1 receptor type I; Interleukin-1 receptor alpha; CD121 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110859 325.00
Check your price
  • IL1R1 Monoclonal Antibody for IHC, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110857
  • Size 200µg
Applications
ELISA (5 µg/m), Immunohistochemistry (10-100 µg/ml), Radioimmune Assays (20-30 ng/ml)
Alternate names:
p80; Interleukin-1 receptor type I; Interleukin-1 receptor alpha; CD121 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110857 325.00
Check your price
  • IL18RAP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141323
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Interleukin-18 receptor beta; Interleukin-18 receptor accessory protein-like; Interleukin-1 receptor 7; IL-1R accessory protein-like; CDw218b; CD218 antigen-like family member B; Accessory protein-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141323 309.00
Check your price
  • IRAK2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520163
  • Size 100ug
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
6330415L08Rik, AI649099, IRAK-2; interleukin-1 receptor-associated kinase 2c; interleukin-1 receptor-associated kinase-like 2; mu-IRAK-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520163 349.00
Check your price
  • IRAK2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA520164
  • Size 100ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
6330415L08Rik, AI649099, IRAK-2; interleukin-1 receptor-associated kinase 2c; interleukin-1 receptor-associated kinase-like 2; mu-IRAK-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520164 349.00
Check your price
  • IRAK2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519937
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
6330415L08Rik, AI649099, IRAK-2; interleukin-1 receptor-associated kinase 2c; interleukin-1 receptor-associated kinase-like 2; mu-IRAK-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519937 349.00
Check your price
  • IRAK2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519917
  • Size 100ug
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (10 ug/ml), Western Blot (2 µg/ml)
Alternate names:
IRAK-2; interleukin-1 receptor associated kinase-2; interleukin-1 receptor-associated kinase-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519917 349.00
Check your price
  • Phospho-Il13Ra1 (Tyr405) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA540237
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Novel cytokine receptor 4; Interleukin-13-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540237 309.00
Check your price
  • IL-33 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515773
  • Size 100µg
Applications
Immunohistochemistry (5 µg/ml), Western Blot (1 ug/ml)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515773 349.00
Check your price
  • IL-33 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515772
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515772 349.00
Check your price
  • IL1R2 Polyclonal Antibody for ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA135856
  • Size 100µg
Applications
ELISA (Assay dependent.)
Alternate names:
Interleukin-1 receptor type II; Interleukin-1 receptor beta; IL-1 type II receptor; CDw121b; CD121 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA135856 700.00
Check your price
  • IL-33 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520397
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (20 ug/ml), Western Blot (1-2 µg/ml)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520397 349.00
Check your price
  • IL-33 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520398
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 µg/ml)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520398 349.00
Check your price
  • IL-33 Polyclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547006
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (0.75-3.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547006 375.00
Check your price
  • IL-33 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523673
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523673 349.00
Check your price
  • IL-33 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523609
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523609 289.00
Check your price
  • IL-33 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA523640
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
C9orf26, DVS27, IL1F11, NF-HEV, NFEHEV, RP11-575C20.2, DVS27-related protein, IL-1F11, interleukin-1 family member 11, interleukin-33, nuclear factor for high endothelial venules, nuclear factor from high endothelial venules
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523640 309.00
Check your price
  • IL-33 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547007
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Frozen) (5-15 µg/mL), Neutralization (10-50 ng/ml), Western Blot (0.4 µg/ml)
Alternate names:
C9orf26, DVS27, IL1F11, NF-HEV, NFEHEV, RP11-575C20.2, DVS27-related protein, IL-1F11, interleukin-1 family member 11, interleukin-33, nuclear factor for high endothelial venules, nuclear factor from high endothelial venules
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547007 399.00
Check your price