1 - 30 of 30 products displayed
  • Phospho-GluR2 (Ser880) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538134
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Glutamate receptor ionotropic, AMPA 2; GluR-K2; GluR-B; AMPA-selective glutamate receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538134 309.00
Check your price
  • Phospho-GluR2 (Tyr876) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517096
  • Size 100 µL
Applications
Western Blot (1:500)
Alternate names:
Glutamate receptor ionotropic, AMPA 2; GluR-K2; GluR-B; AMPA-selective glutamate receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517096 309.00
Check your price
  • GluR2 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519496
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (1:300), Western Blot (3-10 µg/ml)
Alternate names:
Glutamate receptor ionotropic, AMPA 2; GluR-K2; GluR-B; AMPA-selective glutamate receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519496 349.00
Check your price
  • GluR2 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # 320300
  • Size 100 µg
Applications
ELISA (0.5-5 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-5 ug/ml)
Alternate names:
Glutamate receptor ionotropic, AMPA 2; GluR-K2; GluR-B; AMPA-selective glutamate receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
320300 307.00
Check your price
  • GluR2/GluR3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538105
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Glutamate receptor ionotropic, AMPA 3; GluR-K3; GluR-C; AMPA-selective glutamate receptor 3; Glutamate receptor ionotropic, AMPA 2; GluR-K2; GluR-B; AMPA-selective glutamate receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538105 309.00
Check your price
  • GluR2/GluR3/GluR4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517314
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Glutamate receptor ionotropic, AMPA 4; GluR-D; AMPA-selective glutamate receptor 4; Glutamate receptor ionotropic, AMPA 3; GluR-K3; GluR-C; AMPA-selective glutamate receptor 3; Glutamate receptor ionotropic, AMPA 2; GluR-K2; GluR-B; AMPA-selective glutamate receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517314 275.00
Check your price
  • mGluR2/mGluR3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536519
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Metabotropic glutamate receptor 3,mGluR3,GRM3,GPRC1C, MGLUR3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536519 309.00
Check your price
  • mGluR5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513361
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Metabotropic glutamate receptor 5 [Precursor]; mGluR5; GRM5; GPRC1E; MGLUR5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513361 349.00
Check your price
  • Phospho-GluR1 (Thr840) Polyclonal Antibody for Western Blot, IF, IHC (F) and IP
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519451
  • Size 100 µg
Applications
Immunofluorescence (1:100), Immunohistochemistry (Frozen) (1:100), Immunoprecipitation (1:100), Western Blot (1:100)
Alternate names:
Glutamate receptor ionotropic, AMPA 1; GluR-K1; GluR-A; AMPA-selective glutamate receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519451 349.00
Check your price
  • GluR2/GluR3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137774
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Glutamate receptor ionotropic, AMPA 2; GluR-K2; GluR-B; AMPA-selective glutamate receptor 2; Glutamate receptor ionotropic, AMPA 3; GluR-K3; GluR-C; AMPA-selective glutamate receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137774 475.00
Check your price
  • GluR2 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517084
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Western Blot (1:500-1:2000)
Alternate names:
Glutamate receptor ionotropic, AMPA 2; GluR-K2; GluR-B; AMPA-selective glutamate receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517084 309.00
Check your price
  • GluR2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14659
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Glutamate receptor ionotropic, AMPA 2; GluR-K2; GluR-B; AMPA-selective glutamate receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14659 375.00
Check your price
  • GluR2/GluR3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14660
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Glutamate receptor ionotropic, AMPA 2; GluR-K2; GluR-B; AMPA-selective glutamate receptor 2; Glutamate receptor ionotropic, AMPA 3; GluR-K3; GluR-C; AMPA-selective glutamate receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14660 375.00
Check your price
  • mGluR2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532860
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (4 µg/ml)
Alternate names:
GLUR2, GPRC1B, MGLUR2, mGlu2, glutamate metabotropic receptor 2, glutamate receptor homolog, metabotropic glutamate receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532860 349.00
Check your price
  • GRID2 Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536405
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Glutamate receptor ionotropic, delta-2, GluD2, GluR delta-2 subunit, GRID2, GLURD2, GluR delta2, GluR delta2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536405 309.00
Check your price
  • Phospho-GluR1 (Ser849) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536848
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Glutamate receptor ionotropic, AMPA 1; GluR-K1; GluR-A; AMPA-selective glutamate receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536848 349.00
Check your price
  • Phospho-GluR1 (Ser863) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536849
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Glutamate receptor ionotropic, AMPA 1; GluR-K1; GluR-A; AMPA-selective glutamate receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536849 349.00
Check your price
  • Phospho-GluR4 (Ser862) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536807
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Glutamate receptor ionotropic, AMPA 4; GluR-D; AMPA-selective glutamate receptor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536807 349.00
Check your price
  • Phospho-GluR2 (Ser880) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # P21949
  • Size 100µL
Applications
Western Blot (Assay Dependent)
Alternate names:
Glutamate receptor ionotropic, AMPA 2; GluR-K2; GluR-B; AMPA-selective glutamate receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
P21949 309.00
Check your price
  • GluR2/GluR3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137773
  • Size 7 mL
Applications
Immunohistochemistry (Paraffin) (Ready-to-use)
Alternate names:
Glutamate receptor ionotropic, AMPA 2; GluR-K2; GluR-B; AMPA-selective glutamate receptor 2; Glutamate receptor ionotropic, AMPA 3; GluR-K3; GluR-C; AMPA-selective glutamate receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137773 349.00
Check your price
  • mGluR2/mGluR3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Avian, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA121034
  • Size 7 mL
Applications
Immunohistochemistry (Paraffin) (Ready-to-use)
Alternate names:
MGLUR2, mGlu2, Metabotropic glutamate receptor 2, Metabotropic glutamate receptor 3, GPRC1B, GRM2, GRM3, GPRC1C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121034 399.00
Check your price
  • Phospho-GluR1 (Ser849) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537588
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Glutamate receptor ionotropic, AMPA 1; GluR-K1; GluR-A; AMPA-selective glutamate receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537588 309.00
Check your price
  • NMDAR2D Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536539
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200)
Alternate names:
N-methyl D-aspartate receptor subtype 2D; Glutamate [NMDA] receptor subunit epsilon-4; EB11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536539 309.00
Check your price
  • mGluR2/mGluR3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532426
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
GLUR3, GPRC1C, MGLUR3, mGlu3, glutamate metabotropic receptor 3, metabotropic glutamate receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532426 349.00
Check your price
  • NMDAR2D Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA539190
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
N-methyl D-aspartate receptor subtype 2D; Glutamate [NMDA] receptor subunit epsilon-4; EB11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539190 309.00
Check your price
  • Phospho-GluR1 (Ser831) Polyclonal Antibody for Western Blot, IF, IHC and ELISA
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # 368200
  • Size 150 µl
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay-Dependent)
Alternate names:
Glutamate receptor ionotropic, AMPA 1; GluR-K1; GluR-A; AMPA-selective glutamate receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
368200 304.00
Check your price
  • Phospho-GluR1 (Ser845) Polyclonal Antibody for Western Blot, IF, IHC and ELISA
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # 368300
  • Size 150 µl
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay-Dependent)
Alternate names:
Glutamate receptor ionotropic, AMPA 1; GluR-K1; GluR-A; AMPA-selective glutamate receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
368300 309.00
Check your price
  • mGluR2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123573
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123573 375.00
Check your price
  • mGluR2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519348
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (3-5 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
GRM2; glutamate receptor, metabotropic 2; GLUR2; GPRC1B; MGLUR2; mGlu2; glutamate metabotropic receptor 2; glutamate receptor homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519348 349.00
Check your price
  • mGluR2/mGluR3 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA112754
  • Size 10 Miniblots
Applications
Immunohistochemistry (Frozen) (1:500), Western Blot (1:1000)
Alternate names:
MGLUR2, mGlu2, Metabotropic glutamate receptor 2, Metabotropic glutamate receptor 3, GPRC1B, GRM2, GRM3, GPRC1C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112754 425.00
Check your price