1 - 30 of 30 products displayed
  • CXCL10 Polyclonal Antibody for Western Blot, IF, ICC, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546999
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (1-4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine B10; 10 kDa interferon gamma-induced protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546999 375.00
Check your price
  • CXCL11 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523761
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.2-0.8 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine B11; Interferon-inducible T-cell alpha chemoattractant; Interferon gamma-inducible protein 9; H174; Beta-R1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523761 349.00
Check your price
  • NOP58 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # MA191625
  • Size 250µL
Applications
Immunocytochemistry (1:1000-1:10000), Immunofluorescence (1:1000-1:10,000), Immunoprecipitation (2-4 µl lysate), Western Blot (1:10,000)
Alternate names:
Nucleolar protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191625 349.00
Check your price
  • Caspase 8 Monoclonal Antibody for Western Blot, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511558
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunoprecipitation (2 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
MORT1-associated ced-3 homolog; ICE-like apoptotic protease 5; FADD-like ICE; FADD-homologous ICE/ced-3-like protease; CAP4; Apoptotic protease Mch-5; Apoptotic cysteine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511558 425.00
Check your price
  • ErbB4 Monoclonal Antibody, Biotin conjugate for Western Blot, Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA512885
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1:50-1:500)
Alternate names:
p180erbB4; Tyrosine kinase-type cell surface receptor HER4; Proto-oncogene-like protein c-ErbB-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512885 399.00
Check your price
  • CD47 Monoclonal Antibody for Western Blot, IF, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511895
  • Size 500µL
Applications
Flow Cytometry (0.5 ug/test), Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1:10-1:100)
Alternate names:
Protein MER6; Integrin-associated protein; Antigenic surface determinant protein OA3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511895 309.00
Check your price
  • CD2 Monoclonal Antibody for Western Blot, Flow, IP and IA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA0200
  • Size 200ug
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunoprecipitation (Assay Dependent), Inhibition Assays (0.5 ug/test), Western Blot (0.5-2 µg/ml)
Alternate names:
T-cell surface antigen T11/Leu-5; Rosette receptor; LFA-3 receptor; LFA-2; Erythrocyte receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA0200 349.00
Check your price
  • Cyclin B1 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA514324
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1:100-1:1000)
Alternate names:
G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1,OTTHUMP00000223023
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514324 375.00
Check your price
  • Follicle Stimulating Hormone Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514726
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Follitropin beta chain; Follicle-stimulating hormone beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514726 349.00
Check your price
  • alpha-1 Antitrypsin Monoclonal Antibody for Western Blot, Flow, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514661
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514661 349.00
Check your price
  • p53 Monoclonal Antibody for Western Blot, Flow, IP and ChIP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511296
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Tumor suppressor p53; Phosphoprotein p53; Antigen NY-CO-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511296 309.00
Check your price
  • INSR alpha Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513756
  • Size 500µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunoprecipitation (2 µg/ml), Western Blot (1:100-1:1000)
Alternate names:
CD220, HHF5, IR, IR alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513756 375.00
Check your price
  • Rb Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # MA511384
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
pp110; pRb; p105-Rb; pp105
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511384 375.00
Check your price
  • TAP Tag Polyclonal Antibody for Western Blot, IP and ChIP
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # CAB1001
  • Size 50µg
Applications
ChIP assay (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (0.5 µg/ml)
Alternate names:
tandem affinity purification tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
CAB1001 249.00
Check your price
  • Fibronectin Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512314
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1:10-1:100)
Alternate names:
Cold-insoluble globulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512314 309.00
Check your price
  • beta-1,2 Adaptin Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Bovine, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA125065
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:200)
Alternate names:
Plasma membrane adaptor HA2/AP2 adaptin beta subunit; Clathrin assembly protein complex 2 beta large chain; Beta-adaptin; Beta-2-adaptin; Adaptor-related protein complex 2 subunit beta; Adaptor protein complex AP-2 subunit beta; AP105B; Golgi adaptor HA1/AP1 adaptin beta subunit; Clathrin assembly protein complex 1 beta large chain; Beta-adaptin 1; Beta-1-adaptin; Adaptor-related protein complex 1 subunit beta-1; Adaptor protein complex AP-1 subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125065 349.00
Check your price
  • CES1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524244
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (2-10 µg/ml)
Alternate names:
Triacylglycerol hydrolase; Serine esterase 1; Retinyl ester hydrolase; Monocyte/macrophage serine esterase; Methylumbelliferyl-acetate deacetylase 1; HMSE; Egasyn; Cocaine carboxylesterase; Carboxylesterase 1; Brain carboxylesterase hBr1; Acyl-coenzyme A:cholesterol acyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524244 275.00
Check your price
  • BRG1 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 730011
  • Size 100µg
Applications
Immunofluorescence (1:250), Immunoprecipitation (2 ug), Western Blot (1:2000)
Alternate names:
SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; SNF2-beta; Protein brahma homolog 1; BRG1-associated factor 190A; ATP-dependent helicase SMARCA4; Protein BRG-1; Mitotic growth and transcription activator
Catalog # Size Price (AUD) Your Price (AUD) Qty
730011 309.00
Check your price
  • ENPP5 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524212
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
NPP5, EC 3.1, ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative function), ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative), ectonucleotide pyrophosphatase/phosphodiesterase family member 5, E-NPP 5, KIAA0879, NPP-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524212 275.00
Check your price
  • PRSS3 Polyclonal Antibody for Western Blot, IHC (F) and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA548034
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
MTG, PRSS4, T9, TRY3, TRY4, brain trypsinogen, mesotrypsin, mesotrypsinogen, pancreatic trypsinogen III, protease, serine, 4 (trypsin 4, brain), trypsin III, trypsin IV, trypsin-3, trypsinogen 4, trypsinogen 5, trypsinogen IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548034 309.00
Check your price
  • Serpina12 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547725
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
OL-64, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 12, serpin A12, vaspin, visceral adipose tissue-derived serine protease inhibitor, visceral adipose-specific SERPIN
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547725 349.00
Check your price
  • NAALADL2 Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547647
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
NAALADL2, inactive N-acetylated-alpha-linked acidic dipeptidase-like protein 2, NAALADase L2, N-acetylated alpha-linked acidic dipeptidase 2, N-acetylated alpha-linked acidic dipeptidase-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547647 349.00
Check your price
  • SMARCA2 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 730025
  • Size 100µg
Applications
Immunofluorescence (1:250), Immunoprecipitation (Assay Dependent), Western Blot (1:2000)
Alternate names:
SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 2; SNF2-alpha; Protein brahma homolog; BRG1-associated factor 190B; ATP-dependent helicase SMARCA2
Catalog # Size Price (AUD) Your Price (AUD) Qty
730025 309.00
Check your price
  • Actin Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Non-human primate, Plant, Pig, Protozoa, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA511866
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (2 ug), Western Blot (1:5000-1:20,000)
Alternate names:
Smooth muscle gamma-actin; Gamma-2-actin; Alpha-actin-3; Beta-actin; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1; Alpha-cardiac actin; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511866 399.00
Check your price
  • GASP1 Polyclonal Antibody for Western Blot, IP and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547248
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Neutralization (18.9 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
WFIKKN-related protein; WAP, follistatin, immunoglobulin, Kunitz and NTR domain-containing-related protein; Growth and differentiation factor-associated serum protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547248 349.00
Check your price
  • SRC1 Monoclonal Antibody for Western Blot, IHC (P), Flow, IP and ChIP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA1840
  • Size 100µL
Applications
ChIP assay (5µl per 10^6 cells), Flow Cytometry (1µg per 10^6 cells), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:5000)
Alternate names:
Steroid receptor coactivator 1; Renal carcinoma antigen NY-REN-52; RIP160; Protein Hin-2; Class E basic helix-loop-helix protein 74; Nuclear receptor coactivator protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1840 275.00
Check your price
  • XPNPEP2 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547333
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
APP2, X-Pro aminopeptidase 2, aminoacylproline aminopeptidase, mAmP, membrane-bound APP, membrane-bound AmP, membrane-bound aminopeptidase P, xaa-Pro aminopeptidase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547333 349.00
Check your price
  • TRIM28 Monoclonal Antibody for Western Blot, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 730029
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent)
Alternate names:
Tripartite motif-containing protein 28; RING finger protein 96; Nuclear corepressor KAP-1; KRAB-interacting protein 1; KRAB-associated protein 1; E3 SUMO-protein ligase TRIM28
Catalog # Size Price (AUD) Your Price (AUD) Qty
730029 309.00
Check your price
  • Kininogen Monoclonal Antibody for Western Blot and IP
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523989
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
BDK, KNG, HMWK, alpha-2-thiol proteinase inhibitor, bradykinin, fitzgerald factor, high molecular weight kininogen, kininogen-1, williams-Fitzgerald-Flaujeac factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523989 275.00
Check your price
  • Kininogen Monoclonal Antibody for Western Blot and IP
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523988
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
BDK, KNG, HMWK, alpha-2-thiol proteinase inhibitor, bradykinin, fitzgerald factor, high molecular weight kininogen, kininogen-1, williams-Fitzgerald-Flaujeac factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523988 275.00
Check your price