1 - 2 of 2 products displayed
  • WNT2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534320
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
Int-1-related protein; Int-1-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534320 349.00
Check your price
  • WNT2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534321
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10-15 µg/ml)
Alternate names:
Int-1-related protein; Int-1-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534321 349.00
Check your price