1 - 30 of 30 products displayed
  • Phospho-IRS2 (Ser731) Polyclonal Antibody for Western Blot, IP and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130707
  • Size 50 µg
Applications
ELISA (0.1-1 µg/ml), Immunoprecipitation (3-5 µg x 10^7 cells), Western Blot (0.5-2 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130707 425.00
Check your price
  • IRS2 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517056
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
4PS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517056 275.00
Check your price
  • IRS2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514897
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Insulin receptor substrate 2; IRS-2; IRS2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514897 349.00
Check your price
  • IRS1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520137
  • Size 100 ug
Applications
Immunocytochemistry (2 ug/ml), Immunofluorescence (2 ug/ml), Western Blot (1-4 ug/ml)
Alternate names:
HIRS-1; IRS-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520137 349.00
Check your price
  • SHP-2 Oligoclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710867
  • Size 100 µg
Applications
Flow Cytometry (2-4 µg x 10^6 cells), Western Blot (1- 2 µg/ml)
Alternate names:
SH-PTP3; SH-PTP2; Protein-tyrosine phosphatase 2C; Protein-tyrosine phosphatase 1D
Catalog # Size Price (AUD) Your Price (AUD) Qty
710867 309.00
Check your price
  • IRS1 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11059
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (2 ug/ml)
Alternate names:
Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11059 275.00
Check your price
  • Phospho-IRS1 (Ser307) Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11054
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Phospho-Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11054 349.00
Check your price
  • IRS1 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11057
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11057 275.00
Check your price
  • Phospho-IRS1 (Ser636) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11055
  • Size 100 µg
Applications
Western Blot (2 ug/ml)
Alternate names:
Phospho-Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11055 349.00
Check your price
  • IRS1 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11058
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Western Blot (2 ug/ml)
Alternate names:
Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11058 275.00
Check your price
  • INSR alpha Monoclonal Antibody, Biotin conjugate for IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Sheep, Pig, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA513764
  • Size 500 µL
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (2-4 µg/ml)
Alternate names:
CD220, HHF5, IR, IR alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513764 399.00
Check your price
  • SUMO2/SUMO3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA519418
  • Size 100 µg
Applications
Immunocytochemistry (1:100), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:800), Western Blot (1:1000)
Alternate names:
Ubiquitin-like protein SMT3B; Sentrin-2; SUMO-3; SMT3 homolog 2; HSMT3; Ubiquitin-like protein SMT3A; SMT3 homolog 1; SUMO-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519418 349.00
Check your price
  • Phospho-IRS1 (Tyr1179) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11060
  • Size 100 µg
Applications
Western Blot (Assay dependent)
Alternate names:
Phospho-Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11060 349.00
Check your price
  • Phospho-IR/IGF1R (Tyr1158, Tyr1162, Tyr1163) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44806G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
44806G 304.00
Check your price
  • Phospho-IRS1 (Ser307) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44813G
  • Size 100 µL
Applications
Western Blot (Assay Dependent)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
44813G 309.00
Check your price
  • IRS1 Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA515068
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515068 275.00
Check your price
  • SHP-2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520278
  • Size 100 ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
SH-PTP2; Protein-tyrosine phosphatase SYP; SH-PTP3; Protein-tyrosine phosphatase 2C; Protein-tyrosine phosphatase 1D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520278 349.00
Check your price
  • SHP-2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520279
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
SH-PTP2; Protein-tyrosine phosphatase SYP; SH-PTP3; Protein-tyrosine phosphatase 2C; Protein-tyrosine phosphatase 1D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520279 349.00
Check your price
  • Phospho-ATF2 pSer112 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535739
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
cAMP response element-binding protein CRE-BP1; Histone acetyltransferase ATF2; HB16; Cyclic AMP-responsive element-binding protein 2; Activating transcription factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535739 339.00
Check your price
  • Phospho-ATF2 pSer62 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat, Human, Mouse
  • Conjugate
  • Catalog # PA535740
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
cAMP response element-binding protein CRE-BP1; Histone acetyltransferase ATF2; HB16; Cyclic AMP-responsive element-binding protein 2; Activating transcription factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535740 339.00
Check your price
  • INSR beta Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP, ELISA and AP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA513778
  • Size 500 µL
Applications
Affinity Purification (Assay Dependent), ELISA (Assay Dependent), Flow Cytometry (2 ug/test), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunoprecipitation (2 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
CD220, CD220 beta, HHF5, IR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513778 349.00
Check your price
  • INSR alpha Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow, ELISA and FN
  • Host Mouse
  • Target Species Bovine, Human, Sheep, Pig, Rabbit
  • Conjugate Unconjugated
  • Catalog # AHR0231
  • Size 500 µl
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay-Dependent), Functional Assay (Assay Dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (2-4µg/ml), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (Assay Dependent)
Alternate names:
CD220, CD220 beta, HHF5, IR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHR0231 305.00
Check your price
  • RUNX1 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519638
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (1:500), Western Blot (0.25 µg/ml)
Alternate names:
SL3/AKV core-binding factor alpha B subunit; SL3-3 enhancer factor 1 alpha B subunit; Polyomavirus enhancer-binding protein 2 alpha B subunit; Oncogene AML-1; Core-binding factor subunit alpha-2; Acute myeloid leukemia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519638 375.00
Check your price
  • FRS3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536403
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
FRS2-beta; Suc1-associated neurotrophic factor target 2; FGFR-signaling adaptor SNT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536403 309.00
Check your price
  • EEF2 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA519617
  • Size 100 µg
Applications
Immunofluorescence (5 µg/ml), Western Blot (Assay-Dependent)
Alternate names:
EEF-2, EF2, EF-2; elongation factor 2; polypeptidyl-tRNA translocase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519617 349.00
Check your price
  • DOK4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514755
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Insulin receptor substrate 5; Downstream of tyrosine kinase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514755 349.00
Check your price
  • Phospho-INSR (Tyr1334) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44809G
  • Size 10 blots
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (Assay Dependent)
Alternate names:
CD220, CD220 beta, HHF5, IR beta, Insulin Receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
44809G 304.00
Check your price
  • Phospho-INSR (Tyr972) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44800G
  • Size 10 blots
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (Assay Dependent)
Alternate names:
CD220, CD220 beta, HHF5, IR beta, Insulin Receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
44800G 304.00
Check your price
  • PDGF-BB Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519524
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:500), Western Blot (1:500)
Alternate names:
Proto-oncogene c-Sis; Platelet-derived growth factor beta polypeptide; Platelet-derived growth factor B chain; PDGF-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519524 349.00
Check your price
  • Phospho-GRB10 (Tyr67) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536808
  • Size 100 µl
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Maternally expressed gene 1 protein; GRB10 adapter protein; Insulin receptor-binding protein Grb-IR
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536808 349.00
Check your price