1 - 30 of 30 products displayed
  • IRS1 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700662
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (4-5 ug/ml)
Alternate names:
HIRS-1, Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
700662 309.00
Check your price
  • IRS1 Oligoclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710009
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Western Blot (4-5 ug/ml)
Alternate names:
HIRS-1, Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710009 309.00
Check your price
  • Phospho-IRS1 (Ser323) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540280
  • Size 100 µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
IRS1; Insulin receptor substrate 1; IRS-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540280 309.00
Check your price
  • Phospho-IRS1 (Tyr896) Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44818G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
IRS-1, IRS1, Insulin receptor substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44818G 304.00
Check your price
  • Phospho-IRS1 (Tyr612) Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44816G
  • Size 100 µL
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
IRS-1, IRS1, Insulin receptor substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44816G 304.00
Check your price
  • Phospho-IRS1 (Tyr1179) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44822G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
IRS-1, IRS1, Insulin receptor substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44822G 309.00
Check your price
  • Phospho-IRS1 (Ser312) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44814G
  • Size 100 µL
Applications
Western Blot (Assay Dependent)
Alternate names:
IRS-1, IRS1, Insulin receptor substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44814G 309.00
Check your price
  • Phospho-IRS1 (Ser1101) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536719
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536719 349.00
Check your price
  • Phospho-IRS1 (Ser323) Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536739
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536739 349.00
Check your price
  • Phospho-IRS1 (Ser794) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536752
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536752 349.00
Check your price
  • Phospho-IRS1 (Ser639) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536828
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536828 349.00
Check your price
  • Phospho-IRS1 (Ser794) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA540201
  • Size 100 µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540201 309.00
Check your price
  • Phospho-IRS1 (Ser616) Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44550G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
44550G 305.00
Check your price
  • Phospho-IRS1 (Tyr941) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44820G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
IRS-1, IRS1, Insulin receptor substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44820G 309.00
Check your price
  • Phospho-IRS1 (Ser323) Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA538286
  • Size 100 µg
Applications
Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538286 309.00
Check your price
  • Phospho-IRS1 (Ser794) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538364
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538364 309.00
Check your price
  • Phospho-IRS1 (Ser639) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537613
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537613 309.00
Check your price
  • Phospho-IRS1 (Ser307) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44813G
  • Size 100 µL
Applications
Western Blot (Assay Dependent)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
44813G 309.00
Check your price
  • IRS1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534655
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
HIRS-1, IRS-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534655 349.00
Check your price
  • IRS1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529667
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
HIRS-1, IRS-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529667 349.00
Check your price
  • IRS1 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11059
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (2 ug/ml)
Alternate names:
Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11059 275.00
Check your price
  • IRS1 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11057
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11057 275.00
Check your price
  • IRS1 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11058
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Western Blot (2 ug/ml)
Alternate names:
Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11058 275.00
Check your price
  • IRS1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532455
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
HIRS-1, IRS-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532455 349.00
Check your price
  • IRS1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520137
  • Size 100 ug
Applications
Immunocytochemistry (2 ug/ml), Immunofluorescence (2 ug/ml), Western Blot (1-4 ug/ml)
Alternate names:
HIRS-1; IRS-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520137 349.00
Check your price
  • IRS1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515072
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515072 275.00
Check your price
  • Phospho-IRS1 (Ser307) Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11054
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Phospho-Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11054 349.00
Check your price
  • Phospho-IRS1 (Tyr1179) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11060
  • Size 100 µg
Applications
Western Blot (Assay dependent)
Alternate names:
Phospho-Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11060 349.00
Check your price
  • Phospho-IRS1 (Ser636) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11055
  • Size 100 µg
Applications
Western Blot (2 ug/ml)
Alternate names:
Phospho-Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11055 349.00
Check your price
  • Phospho-IRS1 (Ser312) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126697
  • Size 50 µL
Applications
Western Blot (0.35-1 µg/ml)
Alternate names:
pp185
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126697 349.00
Check your price