1 - 2 of 2 products displayed
  • ITGB1BP3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512391
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Ribosylnicotinic acid kinase 2; Ribosylnicotinamide kinase 2; Nicotinic acid riboside kinase 2; Muscle integrin-binding protein; Integrin beta-1-binding protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512391 349.00
Check your price
  • ITGB1BP3 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA524607
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Ribosylnicotinic acid kinase 2; Ribosylnicotinamide kinase 2; Nicotinic acid riboside kinase 2; Muscle integrin-binding protein; Integrin beta-1-binding protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524607 349.00
Check your price