1 - 30 of 30 products displayed
  • JAB1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529872
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Jun activation domain-binding protein 1; Kip1 C-terminus-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529872 349.00
Check your price
  • JAB1 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 371500
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Jun activation domain-binding protein 1; Kip1 C-terminus-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
371500 309.00
Check your price
  • JAB1 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA125464
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (10-20 µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (0.25-0.5 µg/ml)
Alternate names:
Jun activation domain-binding protein 1; Kip1 C-terminus-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125464 375.00
Check your price
  • JAB1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532456
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Jun activation domain-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532456 349.00
Check your price
  • JAB1 Monoclonal Antibody for Western Blot, IHC and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA123246
  • Size 100 µg
Applications
Immunohistochemistry (Assay-Dependent), Immunoprecipitation (0.5-1 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
Jun activation domain-binding protein 1; Kip1 C-terminus-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123246 309.00
Check your price
  • JAB1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Fruit fly, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA123248
  • Size 100 µL
Applications
Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Jun activation domain-binding protein 1; Kip1 C-terminus-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123248 309.00
Check your price
  • JAB1 Monoclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Mouse
  • Target Species Fruit fly, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA123244
  • Size 100 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Assay dependent), Immunoprecipitation (0.5-1 µg/ml), Western Blot (1:1000)
Alternate names:
Jun activation domain-binding protein 1; Kip1 C-terminus-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123244 375.00
Check your price
  • COPS3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515026
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
JAB1-containing signalosome subunit 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515026 349.00
Check your price
  • GPS1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518160
  • Size 100 ug
Applications
Western Blot (0.5 ug/ml)
Alternate names:
Protein MFH; JAB1-containing signalosome subunit 1; G protein pathway suppressor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518160 349.00
Check your price
  • COPS7A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537150
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
JAB1-containing signalosome subunit 7a; Dermal papilla-derived protein 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537150 309.00
Check your price
  • COPS8 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520539
  • Size 100 ug
Applications
Western Blot (2 µg/ml)
Alternate names:
JAB1-containing signalosome subunit 8; COP9 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520539 349.00
Check your price
  • Phospho-GPS1 (Ser454) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539782
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Protein MFH; JAB1-containing signalosome subunit 1; G protein pathway suppressor 1; MFH protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539782 309.00
Check your price
  • GPS1 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131899
  • Size 100 ug
Applications
ELISA (1:32,000), Western Blot (0.5 µg/ml)
Alternate names:
Protein MFH; JAB1-containing signalosome subunit 1; G protein pathway suppressor 1; MFH protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131899 399.00
Check your price
  • TRIP15 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518161
  • Size 100 ug
Applications
Immunohistochemistry (3-5µg/ml), Western Blot (1-3 ug/ml)
Alternate names:
Thyroid receptor-interacting protein 15; JAB1-containing signalosome subunit 2; Alien homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518161 349.00
Check your price
  • TRIP15 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530309
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Thyroid receptor-interacting protein 15; JAB1-containing signalosome subunit 2; Alien homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530309 349.00
Check your price
  • TRIP15 Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131900
  • Size 100 ug
Applications
ELISA (1:8000), Immunohistochemistry (Paraffin) (3-5 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Thyroid receptor-interacting protein 15; JAB1-containing signalosome subunit 2; Alien homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131900 399.00
Check your price
  • TRIP15 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530723
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
Thyroid receptor-interacting protein 15; JAB1-containing signalosome subunit 2; Alien homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530723 349.00
Check your price
  • TRIP15 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528430
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Thyroid receptor-interacting protein 15; JAB1-containing signalosome subunit 2; Alien homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528430 349.00
Check your price
  • GPS1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527149
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein MFH; JAB1-containing signalosome subunit 1; G protein pathway suppressor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527149 349.00
Check your price
  • COPS6 Monoclonal Antibody for IHC (PFA) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521211
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunohistochemistry (PFA fixed) (3 µg/ml)
Alternate names:
Vpr-interacting protein; MOV34 homolog; JAB1-containing signalosome subunit 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521211 309.00
Check your price
  • COPS6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544972
  • Size 100 µl
Applications
Western Blot (1:200)
Alternate names:
Vpr-interacting protein; MOV34 homolog; JAB1-containing signalosome subunit 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544972 309.00
Check your price
  • COPS3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530275
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
JAB1-containing signalosome subunit 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530275 349.00
Check your price
  • COPS4 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531039
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
JAB1-containing signalosome subunit 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531039 349.00
Check your price
  • COPS4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531125
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
JAB1-containing signalosome subunit 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531125 349.00
Check your price
  • TRIP15 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14139
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Thyroid receptor-interacting protein 15; JAB1-containing signalosome subunit 2; Alien homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14139 309.00
Check your price
  • COPS2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Fruit fly, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522802
  • Size 100 ug
Applications
Western Blot (1:100-1:2000)
Alternate names:
Alien protein; Thyroid receptor-interacting protein 15; JAB1-containing signalosome subunit 2; Alien homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522802 349.00
Check your price
  • USP7 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512002
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ubiquitin-specific-processing protease 7; Ubiquitin thioesterase 7; Herpesvirus-associated ubiquitin-specific protease; Deubiquitinating enzyme 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512002 349.00
Check your price
  • USP16 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA512019
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ubiquitin-specific-processing protease 16; Ubiquitin thioesterase 16; Deubiquitinating enzyme 16; Ubiquitin-processing protease UBP-M
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512019 349.00
Check your price
  • USP12 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512012
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Ubiquitin-specific-processing protease 12; Ubiquitin-hydrolyzing enzyme 1; Ubiquitin thioesterase 12; Deubiquitinating enzyme 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512012 349.00
Check your price
  • USP12 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA512011
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ubiquitin-specific-processing protease 12; Ubiquitin-hydrolyzing enzyme 1; Ubiquitin thioesterase 12; Deubiquitinating enzyme 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512011 349.00
Check your price