1 - 30 of 30 products displayed
  • JARID2 Polyclonal Antibody for IF, ICC, IHC, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Rabbit
  • Conjugate Unconjugated
  • Catalog # PA116916
  • Size 100µL
Applications
ChIP assay (1:10-1:500), Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay-Dependent), Western Blot (2 µg/ml)
Alternate names:
Jumonji/ARID domain-containing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116916 349.00
Check your price
  • JMJD6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511173
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Phosphatidylserine receptor; Peptide-lysine 5-dioxygenase JMJD6; Lysyl-hydroxylase JMJD6; Jumonji domain-containing protein 6; JmjC domain-containing protein 6; Histone arginine demethylase JMJD6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511173 349.00
Check your price
  • JMJD6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Marsupial, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523305
  • Size 100ug
Applications
Western Blot (0.25-1 µg/ml)
Alternate names:
Phosphatidylserine receptor; Peptide-lysine 5-dioxygenase JMJD6; Lysyl-hydroxylase JMJD6; Jumonji domain-containing protein 6; JmjC domain-containing protein 6; Histone arginine demethylase JMJD6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523305 349.00
Check your price
  • JMJD6 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520808
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Phosphatidylserine receptor; Peptide-lysine 5-dioxygenase JMJD6; Lysyl-hydroxylase JMJD6; Jumonji domain-containing protein 6; JmjC domain-containing protein 6; Histone arginine demethylase JMJD6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520808 349.00
Check your price
  • JMJD1A Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520802
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Jumonji domain-containing protein 1A; JmjC domain-containing histone demethylation protein 2A; Zinc finger protein TSGA; Testis-specific gene A protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520802 349.00
Check your price
  • JMJD1B Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520803
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Jumonji domain-containing protein 1B; JmjC domain-containing histone demethylation protein 2B; Nuclear protein 5qNCA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520803 349.00
Check your price
  • JMJD1B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511147
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Nuclear protein 5qNCA; Jumonji domain-containing protein 1B; JmjC domain-containing histone demethylation protein 2B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511147 349.00
Check your price
  • JMJD1A Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515723
  • Size 100µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Jumonji domain-containing protein 1A; JmjC domain-containing histone demethylation protein 2A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515723 349.00
Check your price
  • JMJD1A Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515724
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Jumonji domain-containing protein 1A; JmjC domain-containing histone demethylation protein 2A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515724 349.00
Check your price
  • JMJD2A Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514782
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Jumonji domain-containing protein 2A; JmjC domain-containing histone demethylation protein 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514782 275.00
Check your price
  • JMJD2A Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515678
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Jumonji domain-containing protein 2A; JmjC domain-containing histone demethylation protein 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515678 349.00
Check your price
  • JMJD2A Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520806
  • Size 100ug
Applications
Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Jumonji domain-containing protein 2A; JmjC domain-containing histone demethylation protein 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520806 349.00
Check your price
  • JMJD2A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511181
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Jumonji domain-containing protein 2A; JmjC domain-containing histone demethylation protein 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511181 349.00
Check your price
  • JMJD2B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520815
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Jumonji domain-containing protein 2B; JmjC domain-containing histone demethylation protein 3B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520815 349.00
Check your price
  • JMJD2B Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526399
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Jumonji domain-containing protein 2B; JmjC domain-containing histone demethylation protein 3B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526399 349.00
Check your price
  • JMJD1C Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520804
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Thyroid receptor-interacting protein 8; Jumonji domain-containing protein 1C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520804 349.00
Check your price
  • JMJD5 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520807
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Jumonji domain-containing protein 5; JmjC domain-containing protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520807 349.00
Check your price
  • JMJD4 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA520805
  • Size 100ug
Applications
Immunocytochemistry (5 ug/ml), Immunofluorescence (5 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Jumonji domain-containing protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520805 349.00
Check your price
  • JMJD7 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520809
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Jumonji domain-containing protein 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520809 349.00
Check your price
  • JMJD8 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520810
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Jumonji domain-containing protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520810 349.00
Check your price
  • KDM2A Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511177
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
[Histone-H3]-lysine-36 demethylase 1A; JmjC domain-containing histone demethylation protein 1A; F-box/LRR-repeat protein 11; F-box protein Lilina; F-box protein FBL7; F-box and leucine-rich repeat protein 11; CXXC-type zinc finger protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511177 349.00
Check your price
  • KDM2A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511176
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
[Histone-H3]-lysine-36 demethylase 1A; JmjC domain-containing histone demethylation protein 1A; F-box/LRR-repeat protein 11; F-box protein Lilina; F-box protein FBL7; F-box and leucine-rich repeat protein 11; CXXC-type zinc finger protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511176 349.00
Check your price
  • JMJD6 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523089
  • Size 50µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:2000)
Alternate names:
Phosphatidylserine receptor; Peptide-lysine 5-dioxygenase JMJD6; Lysyl-hydroxylase JMJD6; Jumonji domain-containing protein 6; JmjC domain-containing protein 6; Histone arginine demethylase JMJD6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523089 349.00
Check your price
  • JMJD6 Monoclonal Antibody, Biotin conjugate for IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # 730091
  • Size 100µg
Applications
Immunoprecipitation (0.5 mg/ml)
Alternate names:
Phosphatidylserine receptor; Peptide-lysine 5-dioxygenase JMJD6; Lysyl-hydroxylase JMJD6; Jumonji domain-containing protein 6; JmjC domain-containing protein 6; Histone arginine demethylase JMJD6
Catalog # Size Price (AUD) Your Price (AUD) Qty
730091 309.00
Check your price
  • JMJD2C Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523065
  • Size 100µL
Applications
ChIP assay (Assay-Dependent), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:10,000)
Alternate names:
Jumonji domain-containing protein 2C; JmjC domain-containing histone demethylation protein 3C; Gene amplified in squamous cell carcinoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523065 349.00
Check your price
  • JMJD1B Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA514792
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Nuclear protein 5qNCA; Jumonji domain-containing protein 1B; JmjC domain-containing histone demethylation protein 2B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514792 275.00
Check your price
  • JMJD1B Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517170
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Jumonji domain-containing protein 1B; JmjC domain-containing histone demethylation protein 2B; Nuclear protein 5qNCA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517170 275.00
Check your price
  • JMJD2A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530712
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Jumonji domain-containing protein 2A; JmjC domain-containing histone demethylation protein 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530712 349.00
Check your price
  • JMJD2A Polyclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523067
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:25,000)
Alternate names:
Jumonji domain-containing protein 2A; JmjC domain-containing histone demethylation protein 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523067 349.00
Check your price
  • JMJD2B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA524611
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Jumonji domain-containing protein 2B; JmjC domain-containing histone demethylation protein 3B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524611 349.00
Check your price