1 - 30 of 30 products displayed
  • RANK Monoclonal Antibody for Western Blot, Flow and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA141048
  • Size 100µg
Applications
Flow Cytometry (5 µg/10^6 cells), Immunoprecipitation (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Receptor activator of NF-KB; Osteoclast differentiation factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141048 309.00
Check your price
  • Phospho-IkB alpha (Ser32) Oligoclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # 710295
  • Size 100µg
Applications
Flow Cytometry (1-3µg x 10^6 cells), Western Blot (1-3µg/ml)
Alternate names:
Major histocompatibility complex enhancer-binding protein MAD3; I-kappa-B-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
710295 309.00
Check your price
  • POLL Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526937
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
DNA polymerase kappa; DNA polymerase beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526937 349.00
Check your price
  • Rat anti-Mouse Lambda Light Chain Secondary Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A15380
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Immunoglobulin lambda light chain, Ig lambda light chain, Ig lambda, lambda light chain, Igl
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15380 205.00
Check your price
  • Rat anti-Mouse Lambda Light Chain Secondary Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MLAMBDA01
  • Size 500µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Immunoglobulin lambda light chain, Ig lambda light chain, Ig lambda, lambda light chain, Igl
Catalog # Size Price (AUD) Your Price (AUD) Qty
MLAMBDA01 284.00
Check your price
  • POLL Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518228
  • Size 100ug
Applications
Immunohistochemistry (5-10µg/ml)
Alternate names:
DNA polymerase kappa; DNA polymerase beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518228 349.00
Check your price
  • Rat anti-Mouse Lambda Light Chain Secondary Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MLAMBDA04
  • Size 1mL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Immunoglobulin lambda light chain, Ig lambda light chain, Ig lambda, lambda light chain, Igl
Catalog # Size Price (AUD) Your Price (AUD) Qty
MLAMBDA04 202.00
Check your price
  • Mouse anti-Human IgG Kappa Light Chain Secondary Antibody for Western Blot, IF, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 053900
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunofluorescence (1:500-1:20,000), Immunohistochemistry (Paraffin) (1:500-1:20,000), Immunoprecipitation (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
053900 210.00
Check your price
  • Mouse anti-Human Lambda Light Chain Secondary Antibody for Western Blot, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511590
  • Size 500µL
Applications
ELISA (1ug/ml), Flow Cytometry (1ug/ml), Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (1ug/ml)
Alternate names:
Immunoglobulin lambda light chain, Ig lambda light chain, Ig lambda, lambda light chain, Igl
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511590 249.00
Check your price
  • F(ab')2-Goat anti-Human IgG (Kappa light chain) Secondary Antibody, PE conjugate for Flow
  • Host Goat
  • Target Species Human
  • Conjugate PE
  • Catalog # PA174408
  • Size 500µL
Applications
Flow Cytometry (1:20-1:30)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA174408 294.00
Check your price
  • F(ab')2-Goat anti-Human IgG (Kappa light chain) Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # PA174407
  • Size 1mL
Applications
ELISA (1:4000-1:8000), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA174407 379.00
Check your price
  • Goat anti-Mouse IgG (Kappa light chain) Secondary Antibody, HRP conjugate for Western Blot, IHC (F) and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA186015
  • Size 1mL
Applications
ELISA (1:4000-1:8000), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186015 487.00
Check your price
  • Sheep anti-Human IgG3 Secondary Antibody, HRP conjugate for IHC (F) and ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate HRP
  • Catalog # PA186429
  • Size 1mL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (1:100-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186429 1,082.00
Check your price
  • Mouse anti-Human Kappa Light Chain Secondary Antibody, Biotin conjugate for Western Blot, IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA512114
  • Size 500µL
Applications
ELISA (1 µg/ml), Flow Cytometry (1 µg/ml), Immunohistochemistry (Paraffin) (1:400-1:800), Immunoprecipitation (2ug), Western Blot (1:400-1:1600)
Alternate names:
Immunoglobulin kappa light chain, Ig kappa light chain, Ig kappa, kappa light chain, Igk
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512114 309.00
Check your price
  • Mouse anti-Human Kappa Light Chain Secondary Antibody for Western Blot, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512117
  • Size 500µL
Applications
ELISA (1 µg/ml), Flow Cytometry (1 µg/ml), Immunohistochemistry (Paraffin) (1:400-800), Western Blot (1:400-1:1600)
Alternate names:
Immunoglobulin kappa light chain, Ig kappa light chain, Ig kappa, kappa light chain, Igk
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512117 219.00
Check your price
  • Mouse anti-Human Lambda Light Chain Secondary Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512410
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512410 225.00
Check your price
  • TCR V beta 10b Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A18479
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
TCR Vb10b, TCRV beta-10b
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18479 117.00
Check your price
  • Mouse anti-Human Kappa Light Chain Secondary Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA110388
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110388 249.00
Check your price
  • Mouse anti-Human Kappa Light Chain Secondary Antibody for IHC (F), IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110385
  • Size 100µg
Applications
ELISA (Assay dependent), Flow Cytometry (2-8 ug/ml), Immunohistochemistry (Paraffin, Frozen) (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110385 225.00
Check your price
  • Mouse anti-Human Lambda Light Chain Secondary Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110396
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110396 275.00
Check your price
  • Mouse anti-Human Lambda Light Chain Secondary Antibody for IHC (F), IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110393
  • Size 100ug
Applications
ELISA (Assay-Dependent), Flow Cytometry (2-8 ug/ml), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110393 225.00
Check your price
  • Mouse anti-Human Kappa Light Chain Secondary Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110389
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110389 275.00
Check your price
  • Mouse anti-Human Lambda Light Chain Secondary Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA110395
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110395 249.00
Check your price
  • Goat anti-Mouse IgG Fc Cross-Adsorbed Secondary Antibody, FITC conjugate for IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # 31630
  • Size 1mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31630 156.00
Check your price
  • Rabbit anti-Human IgG Fc gamma Secondary Antibody for ELISA and IE
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # SA1600
  • Size 2mg
Applications
ELISA (Assay dependent), Immunoelectrophoresis (assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA1600 181.00
Check your price
  • Rabbit anti-Human IgM Secondary Antibody for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # SA1400
  • Size 1.5mg
Applications
ELISA (Assay dependent), Immunohistochemistry (1:50-1:100), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA1400 211.00
Check your price
  • Mouse anti-Human Lambda Light Chain Secondary Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA110394
  • Size 100Tests
Applications
Flow Cytometry (10 ul/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110394 275.00
Check your price
  • Goat anti-Human IgG (Lambda light chain) Secondary Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 31131
  • Size 500ug
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31131 194.00
Check your price
  • Goat anti-Human IgG (Kappa light chain) Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # 31780
  • Size 500ug
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31780 205.00
Check your price
  • Goat anti-Human IgG (Kappa light chain) Secondary Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 31129
  • Size 500ug
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31129 195.00
Check your price