1 - 14 of 14 products displayed
  • KARS Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515426
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Lysyl-tRNA synthetase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515426 349.00
Check your price
  • HSD17B12 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526360
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Short chain dehydrogenase/reductase family 12C member 1; Estradiol 17-beta-dehydrogenase 12; 3-ketoacyl-CoA reductase; 17-beta-hydroxysteroid dehydrogenase 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526360 349.00
Check your price
  • KIR2D Monoclonal Antibody for Western Blot, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516887
  • Size 200 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (1:50-1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
KIR2DL, KIR2DS1, KIR2DS2, p50.3, KIR2DS4, CD158a, CD158b, CD158, XXbac-BCX195L8.1, KIR-K64, KIR221, NKAT, NKAT1, p58.1, CD158 antigen-like family member A, MHC class I NK cell receptor, NKAT-1, killer Ig receptor, killer cell immunoglobulin-like receptor 2DL1, CD158B2, KIRCL23, NKAT2, CD158B1, NKAT6, CD158J, NKAT5, CD158I, KKA3, NKAT8, CD158H
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516887 399.00
Check your price
  • KARS Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527620
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Lysyl-tRNA synthetase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527620 349.00
Check your price
  • KARS Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527633
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Lysyl-tRNA synthetase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527633 349.00
Check your price
  • TYROBP Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526861
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Killer-activating receptor-associated protein; DNAX-activation protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526861 349.00
Check your price
  • TYROBP Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA112701
  • Size 100 µg
Applications
Immunoprecipitation (2-4µg/ml protein lysate), Western Blot (Assay Dependent)
Alternate names:
Killer-activating receptor-associated protein; DNAX-activation protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112701 375.00
Check your price
  • KIR2DS4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529577
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
P58 natural killer cell receptor clones CL-39/CL-17; Natural killer-associated transcript 8; MHC class I NK cell receptor; CD158 antigen-like family member I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529577 349.00
Check your price
  • TYROBP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129027
  • Size 100 µg
Applications
Western Blot (1:2000)
Alternate names:
Killer-activating receptor-associated protein; DNAX-activation protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129027 349.00
Check your price
  • TYROBP Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528803
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Killer-activating receptor-associated protein; DNAX-activation protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528803 349.00
Check your price
  • TYROBP Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131292
  • Size 100 µg
Applications
Immunoprecipitation (2-4 µg/mg of protein lysate), Western Blot (Assay dependent)
Alternate names:
Killer-activating receptor-associated protein; DNAX-activation protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131292 449.00
Check your price
  • KIR2D Monoclonal Antibody for Western Blot, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182651
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1/50-1/100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p58 natural killer cell receptor clone CL-49; Natural killer-associated transcript 5; NK receptor 183 ActI; MHC class I NK cell receptor; CD158 antigen-like family member J; P58 natural killer cell receptor clones CL-39/CL-17; Natural killer-associated transcript 8; CD158 antigen-like family member I; p58.1 MHC class-I-specific NK receptor; p58 natural killer cell receptor clones CL-42/47.11; Natural killer-associated transcript 1; CD158 antigen-like family member A; MHC class I NK cell receptor Eb6 ActI; CD158 antigen-like family member H
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182651 349.00
Check your price
  • GRIK4 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520452
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Glutamate receptor KA-1; Excitatory amino acid receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520452 349.00
Check your price
  • GRIK4 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520453
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Glutamate receptor KA-1; Excitatory amino acid receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520453 349.00
Check your price