1 - 30 of 30 products displayed
  • KPNA2 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA127857
  • Size 50µg
Applications
Western Blot (1:2000)
Alternate names:
SRP1-alpha; RAG cohort protein 1; Pore targeting complex 58 kDa subunit; Pendulin; Karyopherin subunit alpha-2; Importin alpha P1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127857 425.00
Check your price
  • KPNA3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530075
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
SRP1-gamma; Karyopherin subunit alpha-3; Importin alpha Q2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530075 349.00
Check your price
  • KPNA3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528415
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
SRP1-gamma; Karyopherin subunit alpha-3; Importin alpha Q2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528415 349.00
Check your price
  • KPNA2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521748
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
SRP1-alpha; RAG cohort protein 1; Karyopherin subunit alpha-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521748 349.00
Check your price
  • KPNA2 Monoclonal Antibody for Western Blot
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517016
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
SRP1-alpha; RAG cohort protein 1; Pore targeting complex 58 kDa subunit; Pendulin; Karyopherin subunit alpha-2; Importin alpha P1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517016 425.00
Check your price
  • KPNA2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521034
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
SRP1-alpha; RAG cohort protein 1; Pore targeting complex 58 kDa subunit; Pendulin; Karyopherin subunit alpha-2; Importin alpha P1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521034 349.00
Check your price
  • KPNA2 Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Goat
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132094
  • Size 100ug
Applications
ELISA (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (0.1-0.5 µg/ml)
Alternate names:
SRP1-alpha; RAG cohort protein 1; Pore targeting complex 58 kDa subunit; Pendulin; Karyopherin subunit alpha-2; Importin alpha P1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132094 399.00
Check your price
  • KPNA2 Polyclonal Antibody for Western Blot, IF, ICC and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518237
  • Size 100ug
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Western Blot (1-3 µg/ml)
Alternate names:
SRP1-alpha; RAG cohort protein 1; Karyopherin subunit alpha-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518237 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511810
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511810 349.00
Check your price
  • KPNA1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521032
  • Size 100µg
Applications
Immunocytochemistry (2.5 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
SRP1-beta; RAG cohort protein 2; Nucleoprotein interactor 1; Karyopherin subunit alpha-1; Importin alpha-S1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521032 349.00
Check your price
  • KPNA3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521035
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
SRP1-gamma; Karyopherin subunit alpha-3; Importin alpha Q2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521035 349.00
Check your price
  • KPNA3 Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132096
  • Size 100ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (3-5 µg/ml), Western Blot (0.1-0.5 µg/ml)
Alternate names:
SRP1-gamma; Karyopherin subunit alpha-3; Importin alpha Q2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132096 375.00
Check your price
  • KPNA3 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), IP and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518238
  • Size 100ug
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (5-10µg/ml), Immunoprecipitation (Assay-dependent), Western Blot (0.1-0.5 ug/ml)
Alternate names:
SRP1-gamma; Karyopherin subunit alpha-3; Importin alpha Q2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518238 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518239
  • Size 100ug
Applications
Immunohistochemistry (3µg/ml), Western Blot (0.2-1 ug/ml)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518239 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521033
  • Size 100ug
Applications
Immunocytochemistry (2.5 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521033 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132097
  • Size 100µg
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (0.2-1 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132097 349.00
Check your price
  • KPNA6 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521036
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Karyopherin subunit alpha-6; Importin alpha-S2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521036 349.00
Check your price
  • KPNA5 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529460
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Karyopherin subunit alpha-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529460 349.00
Check your price
  • KPNA5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544199
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Karyopherin subunit alpha-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544199 309.00
Check your price
  • KPNA5 Polyclonal Antibody for IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534371
  • Size 100ug
Applications
Immunohistochemistry (5 ug/mL)
Alternate names:
Karyopherin subunit alpha-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534371 349.00
Check your price
  • KPNA6 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518935
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Karyopherin subunit alpha-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518935 349.00
Check your price
  • KPNA7 Polyclonal Antibody for IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534372
  • Size 100ug
Applications
Immunohistochemistry (5 ug/mL)
Alternate names:
Karyopherin subunit alpha-7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534372 349.00
Check your price
  • KPNA1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529474
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
SRP1-beta; RAG cohort protein 2; Nucleoprotein interactor 1; Karyopherin subunit alpha-1; Importin alpha-S1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529474 349.00
Check your price
  • KPNA1 Monoclonal Antibody for Western Blot, IF, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 370800
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
SRP1-beta; RAG cohort protein 2; Nucleoprotein interactor 1; Karyopherin subunit alpha-1; Importin alpha-S1
Catalog # Size Price (AUD) Your Price (AUD) Qty
370800 303.00
Check your price
  • KPNA1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518236
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
SRP1-beta; RAG cohort protein 2; Nucleoprotein interactor 1; Karyopherin subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518236 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521749
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521749 349.00
Check your price
  • KPNB1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518450
  • Size 100ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Pore targeting complex 97 kDa subunit; Nuclear factor p97; Karyopherin subunit beta-1; Importin-90
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518450 349.00
Check your price
  • TNPO1 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524581
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
M9 region interaction protein; Karyopherin beta-2; Importin beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524581 349.00
Check your price
  • IPO5 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530076
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Ran-binding protein 5; Karyopherin beta-3; Importin subunit beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530076 349.00
Check your price
  • TNPO1 Monoclonal Antibody for Western Blot, IF, IP and ELISA
  • Host Mouse
  • Target Species Human, Non-human primate, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA15885
  • Size 50µL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
M9 region interaction protein; Karyopherin beta-2; Importin beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15885 375.00
Check your price