1 - 5 of 5 products displayed
  • KCNIP2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA541075
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.06 µg/ml)
Alternate names:
Potassium channel-interacting protein 2; A-type potassium channel modulatory protein 2; Cardiac voltage-gated potassium channel modulatory subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541075 309.00
Check your price
  • KCNIP2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA1927
  • Size 100µg
Applications
Western Blot (2 µg/ml)
Alternate names:
Potassium channel-interacting protein 2; A-type potassium channel modulatory protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1927 275.00
Check your price
  • KCNIP1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA141705
  • Size 100µg
Applications
Western Blot (1:100-1:2000)
Alternate names:
Vesicle APC-binding protein; Potassium channel-interacting protein 1; A-type potassium channel modulatory protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141705 309.00
Check your price
  • KCNIP3 Polyclonal Antibody for IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Rabbit
  • Conjugate Unconjugated
  • Catalog # PA1928
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (1:25)
Alternate names:
Potassium Channel Interacting Protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1928 275.00
Check your price
  • Mouse KChlP2 (residues 6-24) Synthetic Peptide for Ctrl
  • Host N/A
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # PEP160
  • Size 50µg
Applications
Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP160 109.00
Check your price