1 - 3 of 3 products displayed
  • KCNE2 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19547
  • Size 500ug
Applications
Western Blot (1:1000)
Alternate names:
Potassium channel subunit beta MiRP1; Minimum potassium ion channel-related peptide 1; MinK-related peptide 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19547 159.00
Check your price
  • HCN1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00122W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Hyperpolarization-activated cation channel 2; Brain cyclic nucleotide-gated channel 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00122W 309.00
Check your price
  • HCN1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSR00123W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Brain cyclic nucleotide-gated channel 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00123W 309.00
Check your price