1 - 3 of 3 products displayed
  • KCNJ3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533868
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (10-15 µg/ml)
Alternate names:
Potassium channel, inwardly rectifying subfamily J member 3; Inward rectifier K(+) channel Kir3.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533868 349.00
Check your price
  • KCNJ3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521975
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Potassium channel, inwardly rectifying subfamily J member 3; Inward rectifier K(+) channel Kir3.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521975 349.00
Check your price
  • Phospho-KCNJ3 (Ser185) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538572
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Potassium channel, inwardly rectifying subfamily J member 3; KGB1; KGA; Inward rectifier K(+) channel Kir3.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538572 309.00
Check your price