1 - 19 of 19 products displayed
  • KDEL Receptor Pan Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Pig, Rabbit, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA190944
  • Size 50 µL
Applications
Immunocytochemistry (1:900-1:1100), Immunofluorescence (1:900-1:1100), Immunoprecipitation (1:400-1:600), Western Blot (1:200-1:300)
Alternate names:
KDEL endoplasmic reticulum protein retention receptor 2; Putative MAPK-activating protein PM23; KDEL endoplasmic reticulum protein retention receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190944 425.00
Check your price
  • KDELR2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539163
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
KDEL endoplasmic reticulum protein retention receptor 2; ERD2-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539163 309.00
Check your price
  • KDELR3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Human
  • Conjugate Unconjugated
  • Catalog # PA539164
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
KDEL endoplasmic reticulum protein retention receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539164 309.00
Check your price
  • Calreticulin Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Mouse
  • Target Species Hamster, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA191034
  • Size 100 µg
Applications
Flow Cytometry (1:100), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (2 µg/ml), Immunoprecipitation (12.5 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
grp60; HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191034 475.00
Check your price
  • Calreticulin Polyclonal Antibody for Western Blot and IHC (F)
  • Host Goat
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA133045
  • Size 50 µL
Applications
Immunohistochemistry (Frozen) (1:50-1:70), Western Blot (1:300-1:500)
Alternate names:
HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55; Calcium-binding protein 3; CALBP; grp60
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA133045 449.00
Check your price
  • KDEL Polyclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1013
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (1:100-200), Immunoprecipitation (Assay dependent), Western Blot (8-16 µg/ml)
Alternate names:
Heat shock protein 90 kDa beta member 1; 94 kDa glucose-regulated protein; Steroidogenesis-activator polypeptide; Immunoglobulin heavy chain-binding protein; Heat shock 70 kDa protein 5; gp96 homolog; Tumor rejection antigen 1; Tumor rejection antigen gp96; Polymorphic tumor rejection antigen 1; Endoplasmic reticulum resident protein 99; Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1013 349.00
Check your price
  • GRP78 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1014A
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Western Blot (2 µg/ml)
Alternate names:
Steroidogenesis-activator polypeptide; Immunoglobulin heavy chain-binding protein; Heat shock 70 kDa protein 5; Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1014A 349.00
Check your price
  • PDI Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow, IP and IM
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA3019
  • Size 100 µL
Applications
Flow Cytometry (0.5-1 ug/test), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin, Frozen) (1:100), Immunomicroscopy (Assay Dependent), Immunoprecipitation (Assay dependent), Western Blot (1:200-1:2000)
Alternate names:
Prolyl 4-hydroxylase subunit beta; Cellular thyroid hormone-binding protein; p55; Endoplasmic reticulum resident protein 59
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3019 375.00
Check your price
  • GRP94 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137807
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
gp96 homolog; Tumor rejection antigen 1; Heat shock protein 90 kDa beta member 1; 94 kDa glucose-regulated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137807 425.00
Check your price
  • Calreticulin Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA3900
  • Size 100 µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55; Calcium-binding protein 3; CALBP; grp60
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3900 375.00
Check your price
  • Calnexin Monoclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA3027
  • Size 100 µL
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (1:20), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:1000)
Alternate names:
p90; Major histocompatibility complex class I antigen-binding protein p88; IP90
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3027 375.00
Check your price
  • GRP94 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rat
  • Target Species Rat, Chicken, Hamster, Human, Mouse, Bovine
  • Conjugate Unconjugated
  • Catalog # MA3016
  • Size 100 µL
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:1000)
Alternate names:
Heat shock protein 90 kDa beta member 1; 94 kDa glucose-regulated protein; gp96 homolog; Tumor rejection antigen 1; Tumor rejection antigen gp96; Polymorphic tumor rejection antigen 1; Endoplasmic reticulum resident protein 99; Transferrin-binding protein; Heat shock 108 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3016 349.00
Check your price
  • PDI Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Mouse
  • Target Species Dog, Hamster, Human, Mouse, Pig, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA3018
  • Size 100 µL
Applications
Flow Cytometry (Assay dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (1:100), Immunoprecipitation (Assay dependent), Western Blot (1:200-1:2000)
Alternate names:
Prolyl 4-hydroxylase subunit beta; Cellular thyroid hormone-binding protein; p55; Endoplasmic reticulum resident protein 59
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3018 375.00
Check your price
  • Calreticulin Polyclonal Antibody for Western Blot, IF and ICC
  • Host Chicken
  • Target Species Dog, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1902A
  • Size 100 µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Western Blot (1:1000)
Alternate names:
HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55; Calcium-binding protein 3; CALBP; grp60
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1902A 349.00
Check your price
  • Calreticulin Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Chicken
  • Target Species Dog, Hamster, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA1903
  • Size 100 µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55; Calcium-binding protein 3; CALBP; grp60
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1903 375.00
Check your price
  • PDIA6 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA3008
  • Size 100 µL
Applications
Western Blot (1:1,000)
Alternate names:
Thioredoxin domain-containing protein 7; Protein disulfide isomerase P5; Endoplasmic reticulum protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3008 349.00
Check your price
  • Calreticulin Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515382
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55; grp60
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515382 349.00
Check your price
  • Calreticulin Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519854
  • Size 100 µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
grp60; HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519854 349.00
Check your price
  • Calreticulin Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525922
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55; grp60
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525922 349.00
Check your price