1 - 5 of 5 products displayed
  • JMJD2A Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530712
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Jumonji domain-containing protein 2A; JmjC domain-containing histone demethylation protein 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530712 349.00
Check your price
  • JMJD2A Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511181
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Jumonji domain-containing protein 2A; JmjC domain-containing histone demethylation protein 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511181 349.00
Check your price
  • JMJD2A Antibody (Monoclonal, 5H1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515678
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Jumonji domain-containing protein 2A; JmjC domain-containing histone demethylation protein 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515678 349.00
Check your price
  • JMJD2A Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523067
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:25,000)
Alternate names:
Jumonji domain-containing protein 2A; JmjC domain-containing histone demethylation protein 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523067 349.00
Check your price
  • JMJD2A Antibody (Monoclonal, C.37.0)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514782
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Jumonji domain-containing protein 2A; JmjC domain-containing histone demethylation protein 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514782 275.00
Check your price