1 - 30 of 30 products displayed
  • Cytokeratin 10 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532459
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Keratin-10; Cytokeratin-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532459 349.00
Check your price
  • Cytokeratin 10 Monoclonal Antibody for Western Blot, IF, IHC (P) and Flow
  • Host Mouse
  • Target Species Dog, Cat, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA513705
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (0.5-1.0 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Keratin-10; Cytokeratin-10; Type I keratin Ka10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513705 275.00
Check your price
  • Cytokeratin 10 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Dog, Human, Mouse, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA106319
  • Size 100µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1:100-1:1000)
Alternate names:
Keratin-10; Cytokeratin-10; Keratin, type I cytoskeletal 59 kDa; 56 kDa cytokeratin; Type I keratin Ka10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106319 349.00
Check your price
  • Cytokeratin 10 Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Dog, Cat, Human, Rat
  • Conjugate Biotin
  • Catalog # MA513702
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (0.5-1.0 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Keratin-10; Cytokeratin-10; Type I keratin Ka10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513702 349.00
Check your price
  • Cytokeratin 10 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 395300
  • Size 100µg
Applications
Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Keratin-10; Cytokeratin-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
395300 309.00
Check your price
  • Cytokeratin 10 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA15766
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1:1000)
Alternate names:
Keratin-10; Cytokeratin-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15766 425.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Amphibian, Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA512231
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1:10-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512231 225.00
Check your price
  • Cytokeratin Pan Type I/II Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA513156
  • Size 500µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513156 219.00
Check your price
  • Cytokeratin Pan Type I/II Monoclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Horse, Human, Mouse, Non-human primate, Pig, Shrew
  • Conjugate Unconjugated
  • Catalog # MA182041
  • Size 100µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1/10-1/40), Western Blot (1:200-1:1000)
Alternate names:
CK5, DDD, EBS2, K5, KRT5A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182041 159.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA191326
  • Size 500µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:50-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:400)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191326 309.00
Check your price
  • Cytokeratin 10 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513663
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Keratin-10; Cytokeratin-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513663 349.00
Check your price
  • Cytokeratin 10 Monoclonal Antibody for Western Blot and IHC (F)
  • Host Mouse
  • Target Species Bovine, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA135857
  • Size 1mL
Applications
Immunohistochemistry (Frozen) (1:5-1:10), Western Blot (Neat)
Alternate names:
Keratin-10; Cytokeratin-6B; Cytokeratin-10; Cytokeratin VIB; Type I keratin Ka10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135857 399.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA513203
  • Size 500µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1-2 ug/ml), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513203 219.00
Check your price
  • Cytokeratin 18 Monoclonal Antibody for Western Blot, ICC, IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119041
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (10 µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (1-2 µg/ml)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119041 309.00
Check your price
  • Cytokeratin Pan Type I Monoclonal Antibody for Western Blot, IHC (P) and IM
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA15761
  • Size 200µL
Applications
Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay Dependent), Western Blot (1:1000-1:3000)
Alternate names:
Keratin-14; Cytokeratin-14; Keratin-10; Cytokeratin-10; Keratin-15; Cytokeratin-15; Keratin-16; Cytokeratin-16; Keratin-19; Cytokeratin-19
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15761 425.00
Check your price
  • Cytokeratin 5/14 Monoclonal Antibody for Western Blot and IHC (F)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112590
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Keratin-14; Cytokeratin-14; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112590 375.00
Check your price
  • FGF10 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA515192
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Keratinocyte growth factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515192 349.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IHC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112594
  • Size 100µg
Applications
Immunohistochemistry (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p53-related protein; p53-like transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112594 375.00
Check your price
  • Vimentin Monoclonal Antibody for Western Blot, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Mouse, Pig, Rat, Chicken, Human
  • Conjugate Unconjugated
  • Catalog # MA110459
  • Size 100ug
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (5 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
VIM, FLJ36605
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110459 225.00
Check your price
  • Cytokeratin Pan Type II Monoclonal Antibody for Western Blot, IF, IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA513150
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
CK-13, cytokeratin 13, cytokeratin-13, keratin, type I cytoskeletal 13, keratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513150 225.00
Check your price
  • Cytokeratin 14 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA511599
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:800-1600), Western Blot (1-2 µg/ml)
Alternate names:
Keratin-14; Cytokeratin-14; Type I keratin Ka14
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511599 269.00
Check your price
  • Cytokeratin 17 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532461
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Keratin-17; Cytokeratin-17; 39.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532461 349.00
Check your price
  • Cytokeratin 14 Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516370
  • Size 500µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
Keratin-14; Cytokeratin-14
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516370 499.00
Check your price
  • Cytokeratin 14 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516722
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:400-1:800), Western Blot (1 µg/mL)
Alternate names:
Keratin-14; Cytokeratin-14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516722 275.00
Check your price
  • Cytokeratin 14 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532460
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Keratin-14; Cytokeratin-14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532460 225.00
Check your price
  • Cytokeratin 6C Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516373
  • Size 500µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Type-II keratin Kb12; Keratin-6C; Keratin K6h; Cytokeratin-6E; Cytokeratin-6C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516373 499.00
Check your price
  • Cytokeratin 6C Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532466
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Type-II keratin Kb12; Keratin-6C; Keratin K6h; Cytokeratin-6E; Cytokeratin-6C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532466 349.00
Check your price
  • Cytokeratin 18 Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516371
  • Size 500µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516371 499.00
Check your price
  • Cytokeratin 18 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532462
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532462 349.00
Check your price
  • Cytokeratin 8 Monoclonal Antibody for IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514428
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (0.5-1.0 µg/ml), Immunoprecipitation (2µg/mg protein lysate)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514428 309.00
Check your price