1 - 3 of 3 products displayed
  • Cytokeratin 17 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532461
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Keratin-17; Cytokeratin-17; 39.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532461 349.00
Check your price
  • Cytokeratin 17 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA527949
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
Type I keratin Ka17; Keratin-17; Cytokeratin-17; 39.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527949 349.00
Check your price
  • Cytokeratin 17 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529919
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Keratin-17; Cytokeratin-17; 39.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529919 349.00
Check your price