1 - 30 of 30 products displayed
  • Cytokeratin 8 Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516420
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516420 499.00
Check your price
  • Cytokeratin 8 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532469
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532469 199.00
Check your price
  • Cytokeratin 8 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532468
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532468 275.00
Check your price
  • Cytokeratin 8 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137987
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137987 349.00
Check your price
  • Phospho-Cytokeratin 8 (Ser74) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537623
  • Size 100 µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Western Blot (1:500-1:1000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537623 309.00
Check your price
  • Phospho-Cytokeratin 8 (Ser73) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538072
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Cytokeratin endo A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538072 309.00
Check your price
  • Cytokeratin 8 Monoclonal Antibody, Biotin conjugate for IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA514425
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (0.5-1.0 µg/ml), Immunoprecipitation (2 µg/ml)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514425 349.00
Check your price
  • Cytokeratin 8 Monoclonal Antibody for Western Blot, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA106318
  • Size 100 µL
Applications
Flow Cytometry (Assay dependent), Immunocytochemistry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Cytokeratin endo A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106318 309.00
Check your price
  • Cytokeratin 8 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA106317
  • Size 100 µL
Applications
Flow Cytometry (1:25-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:25-1:200), Immunohistochemistry (Paraffin, Frozen) (1:25-1:200), Western Blot (1:100-1:1000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106317 309.00
Check your price
  • Cytokeratin 8 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514428
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (0.5-1.0 µg/ml), Immunoprecipitation (2µg/mg protein lysate)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514428 309.00
Check your price
  • Cytokeratin 8 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529607
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Cytokeratin endo A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529607 349.00
Check your price
  • Cytokeratin 8 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528985
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Cytokeratin endo A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528985 349.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody, APC conjugate for IF, ICC and Flow
  • Host Mouse
  • Target Species Mammal
  • Conjugate APC
  • Catalog # MA110325
  • Size 100 ug
Applications
Flow Cytometry (5 ug/ml), Immunocytochemistry (1:100-1:400), Immunofluorescence (1:100-1:400)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Keratin-10; Cytokeratin-10; Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin; Type-II keratin Kb6; Keratin-6A; Cytokeratin-6D; Cytokeratin-6A; Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110325 349.00
Check your price
  • Cytokeratin Pan Type II Monoclonal Antibody for Western Blot, IF, IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA513150
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
CK-13, cytokeratin 13, cytokeratin-13, keratin, type I cytoskeletal 13, keratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513150 225.00
Check your price
  • Phospho-Cytokeratin 8 (Ser74) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536799
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Cytokeratin endo A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536799 349.00
Check your price
  • Cytokeratin 8 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534762
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Cytokeratin endo A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534762 349.00
Check your price
  • Cytokeratin 8/18 Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA513144
  • Size 500 µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000-1:5000)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Type-II keratin Kb8; Keratin-8; Cytokeratin-A; Cytokeratin-8; Cytokeratin endo A; Cytokeratin endo B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513144 219.00
Check your price
  • Phospho-Cytokeratin 8 (Ser432) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538464
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Cytokeratin endo A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538464 309.00
Check your price
  • Cytokeratin Pan Type I/II Monoclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Horse, Human, Mouse, Non-human primate, Pig, Shrew
  • Conjugate Unconjugated
  • Catalog # MA182041
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1/10-1/40), Western Blot (1:200-1:1000)
Alternate names:
CK5, DDD, EBS2, K5, KRT5A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182041 159.00
Check your price
  • Cytokeratin Pan Type I/II Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA513156
  • Size 500 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513156 219.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody, Alexa Fluor® 488 conjugate for Flow
  • Host Mouse
  • Target Species Mammal
  • Conjugate Alexa Fluor® 488
  • Catalog # MA518156
  • Size 100 µg
Applications
Flow Cytometry (10ug/ml)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Keratin-10; Cytokeratin-10; Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin; Type-II keratin Kb6; Keratin-6A; Cytokeratin-6D; Cytokeratin-6A; Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518156 309.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Amphibian, Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA512231
  • Size 500 µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1:10-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512231 225.00
Check your price
  • Cytokeratin Pan Type II Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134220
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134220 399.00
Check your price
  • Cytokeratin Pan Type II Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126281
  • Size 6 mL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Neat), Immunohistochemistry (Paraffin) (Neat), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126281 349.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA191326
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:50-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:400)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191326 309.00
Check your price
  • Cytokeratin 5/8 Monoclonal Antibody for Western Blot, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Dog, Cat, Hamster, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA106314
  • Size 100 µL
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin; Cytokeratin endo A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106314 349.00
Check your price
  • Cytokeratin 8 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515460
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515460 349.00
Check your price
  • Cytokeratin 8 Monoclonal Antibody for Western Blot, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Human, Sheep, Pig
  • Conjugate Unconjugated
  • Catalog # MA119035
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-A; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119035 199.00
Check your price
  • Cytokeratin 8 Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514476
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514476 850.00
Check your price
  • Cytokeratin 8 Monoclonal Antibody for IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126238
  • Size 500 µL
Applications
Immunohistochemistry (Frozen) (1:10-25), Immunohistochemistry (Paraffin) (1:10-25)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126238 399.00
Check your price