1 - 30 of 30 products displayed
  • Cytokeratin Pan Type II Antibody (Monoclonal, AE3)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA513150
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
CK-13, cytokeratin 13, cytokeratin-13, keratin, type I cytoskeletal 13, keratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513150 225.00
Check your price
  • Cytokeratin Pan Type I/II Antibody (Cocktail, AE1/AE3)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA513156
  • Size 500 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513156 219.00
Check your price
  • Cytokeratin Pan Type II Antibody (Monoclonal, AE3)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA135368
  • Size 250 µg
Applications
Immunohistochemistry (Paraffin, Frozen) (1:150-1:200), Western Blot (1:100)
Alternate names:
Type-II keratin Kb2; Keratin-2e; Keratin-2 epidermis; Epithelial keratin-2e; Cytokeratin-2e
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135368 375.00
Check your price
  • KRT83 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA544338
  • Size 100 µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Type-II keratin Kb23; Type II hair keratin Hb3; Keratin-83; Hair keratin K2.10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544338 309.00
Check your price
  • Cytokeratin Pan Type I Antibody (Monoclonal, AE1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA15761
  • Size 200 µL
Applications
Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay Dependent), Western Blot (1:1000-1:3000)
Alternate names:
Keratin-14; Cytokeratin-14; Keratin-10; Cytokeratin-10; Keratin-15; Cytokeratin-15; Keratin-16; Cytokeratin-16; Keratin-19; Cytokeratin-19
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15761 425.00
Check your price
  • Cytokeratin 17 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532461
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Keratin-17; Cytokeratin-17; 39.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532461 349.00
Check your price
  • Cytokeratin 14 Antibody (Monoclonal, SP53)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516370
  • Size 500 µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
Keratin-14; Cytokeratin-14
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516370 499.00
Check your price
  • Cytokeratin 14 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138001
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Keratin-14; Cytokeratin-14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138001 349.00
Check your price
  • Cytokeratin 14 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532460
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Keratin-14; Cytokeratin-14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532460 225.00
Check your price
  • Cytokeratin Pan Type I/II Antibody (Cocktail, AE1/AE3)
  • Host Mouse
  • Target Species Horse, Human, Mouse, Non-human primate, Pig, Shrew
  • Conjugate Unconjugated
  • Catalog # MA182041
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1/10-1/40), Western Blot (1:200-1:1000)
Alternate names:
CK5, DDD, EBS2, K5, KRT5A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182041 159.00
Check your price
  • Cytokeratin 6C Antibody (Monoclonal, SP87)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516373
  • Size 500 µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Type-II keratin Kb12; Keratin-6C; Keratin K6h; Cytokeratin-6E; Cytokeratin-6C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516373 499.00
Check your price
  • Cytokeratin 6C Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532466
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Type-II keratin Kb12; Keratin-6C; Keratin K6h; Cytokeratin-6E; Cytokeratin-6C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532466 349.00
Check your price
  • Cytokeratin 18 Antibody (Monoclonal, SP69)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516371
  • Size 500 µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516371 499.00
Check your price
  • Cytokeratin 18 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532462
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532462 349.00
Check your price
  • Cytokeratin 8 Antibody (Monoclonal, M20)
  • Host Mouse
  • Target Species Human, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA106318
  • Size 100 µL
Applications
Flow Cytometry (Assay dependent), Immunocytochemistry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Cytokeratin endo A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106318 309.00
Check your price
  • Cytokeratin 17 Antibody (Monoclonal, E3)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA513539
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:40-1:80)
Alternate names:
Type I keratin Ka17; Keratin-17; Cytokeratin-17; 39.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513539 309.00
Check your price
  • Cytokeratin 14 Antibody, Biotin (Monoclonal, LL002)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Biotin
  • Catalog # MA511596
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:800-1:1600), Western Blot (1-2 µg/ml)
Alternate names:
Keratin-14; Cytokeratin-14; Type I keratin Ka14
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511596 349.00
Check your price
  • Cytokeratin 14 Antibody (Monoclonal, LL002)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA511599
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:800-1600), Western Blot (1-2 µg/ml)
Alternate names:
Keratin-14; Cytokeratin-14; Type I keratin Ka14
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511599 269.00
Check your price
  • Cytokeratin 8 Antibody (Monoclonal, SP102)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516420
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516420 499.00
Check your price
  • Cytokeratin 8 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137987
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137987 349.00
Check your price
  • Cytokeratin 8 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532469
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532469 199.00
Check your price
  • Cytokeratin 8 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532468
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532468 275.00
Check your price
  • Cytokeratin 20 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532464
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Protein IT; Keratin-20; Cytokeratin-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532464 349.00
Check your price
  • Cytokeratin 10 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532459
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Keratin-10; Cytokeratin-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532459 349.00
Check your price
  • Cytokeratin 14 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516722
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:400-1:800), Western Blot (1 µg/mL)
Alternate names:
Keratin-14; Cytokeratin-14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516722 275.00
Check your price
  • Cytokeratin 19 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532463
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Keratin-19; Cytokeratin-19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532463 199.00
Check your price
  • Cytokeratin Pan Antibody, APC (Monoclonal, C-11)
  • Host Mouse
  • Target Species Mammal
  • Conjugate APC
  • Catalog # MA110325
  • Size 100 ug
Applications
Flow Cytometry (5 ug/ml), Immunocytochemistry (1:100-1:400), Immunofluorescence (1:100-1:400)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Keratin-10; Cytokeratin-10; Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin; Type-II keratin Kb6; Keratin-6A; Cytokeratin-6D; Cytokeratin-6A; Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110325 349.00
Check your price
  • Cytokeratin Pan Type I Antibody (Monoclonal, AE1)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA513144
  • Size 500 µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000-1:5000)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Type-II keratin Kb8; Keratin-8; Cytokeratin-A; Cytokeratin-8; Cytokeratin endo A; Cytokeratin endo B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513144 219.00
Check your price
  • Cytokeratin 3 Antibody (Monoclonal, AE5)
  • Host Mouse
  • Target Species Bovine, Human, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA15763
  • Size 100 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1:1000-1:3000)
Alternate names:
Type-II keratin Kb3; Keratin-3; Cytokeratin-3; 65 kDa cytokeratin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15763 600.00
Check your price
  • Hair Cortex Cytokeratin Antibody (Monoclonal, AE13)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA15765
  • Size 100 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunohistochemistry (PFA fixed) (Assay Dependent), Western Blot (1:1000)
Alternate names:
Type I hair keratin Ka36; Keratin-40; Cytokeratin-40
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15765 425.00
Check your price