1 - 30 of 30 products displayed
  • KGF Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132177
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (0.5-1 ug/ml)
Alternate names:
Keratinocyte growth factor; Heparin-binding growth factor 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132177 449.00
Check your price
  • TGF beta-2 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710276
  • Size 100µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Polyergin; Glioblastoma-derived T-cell suppressor factor; Cetermin; BSC-1 cell growth inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
710276 309.00
Check your price
  • KGF Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523702
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Keratinocyte growth factor; Heparin-binding growth factor 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523702 349.00
Check your price
  • FGF10 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA515192
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Keratinocyte growth factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515192 349.00
Check your price
  • FGFR2 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514653
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514653 349.00
Check your price
  • FGFR2 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514652
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514652 349.00
Check your price
  • KGF Monoclonal Antibody for IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # MA524009
  • Size 100µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunohistochemistry (Paraffin) (8-25 µg/ml)
Alternate names:
Keratinocyte growth factor; Heparin-binding growth factor 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524009 275.00
Check your price
  • FGFR2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518914
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518914 349.00
Check your price
  • FGFR2 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514651
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514651 349.00
Check your price
  • FGFR2 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514654
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514654 349.00
Check your price
  • FGFR2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137685
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137685 475.00
Check your price
  • FGFR2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532404
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532404 349.00
Check your price
  • FGF10 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527300
  • Size 100µL
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
Keratinocyte growth factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527300 349.00
Check your price
  • FGFR2 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131990
  • Size 400µl
Applications
ELISA (1:900-1:1100), Flow Cytometry (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (1:50-1:100), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:100-1:500)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131990 449.00
Check your price
  • FGFR2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529426
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529426 349.00
Check your price
  • Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537816
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Proto-oncogene c-Fgr; N-sam; Fms-like tyrosine kinase 2; Basic fibroblast growth factor receptor 1; Keratinocyte growth factor receptor; K-sam; MFR
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537816 309.00
Check your price
  • FGFR2 Polyclonal Antibody for Western Blot, IHC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA124763
  • Size 100µL
Applications
Immunohistochemistry (Assay Dependent), Immunoprecipitation (1:2,000), Western Blot (1:2,000)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124763 309.00
Check your price
  • FGFR2 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA124764
  • Size 50µL
Applications
Immunohistochemistry (Paraffin) (1:1000), Immunoprecipitation (1:1,000), Western Blot (1:2,000)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124764 375.00
Check your price
  • FGF10 Polyclonal Antibody for Western Blot, ELISA, Neu and IA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA125518
  • Size 50µg
Applications
ELISA (0.5 µg/ml), Inhibition Assays (0.20-0.35 ug/ml), Neutralization (0.20-0.35 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Keratinocyte growth factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125518 399.00
Check your price
  • KGF Polyclonal Antibody for Western Blot, ELISA, Neu and IA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124683
  • Size 50µg
Applications
ELISA (0.5-1 µg/ml), Inhibition Assays (0.1-100 ug/ml), Neutralization (0.1-100 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Keratinocyte growth factor; Heparin-binding growth factor 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124683 309.00
Check your price
  • DDX11 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523956
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Keratinocyte growth factor-regulated gene 2 protein; DEAD/H box protein 11; CHL1-related protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523956 349.00
Check your price
  • HBEGF Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542703
  • Size 100µl
Applications
Western Blot (1 µg/ml)
Alternate names:
heparin-binding EGF-like growth factor, DTR, DTS, DTSF, HEGFL
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542703 309.00
Check your price
  • IGFBP6 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518929
  • Size 100ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
IGFBP6 ; insulin-like growth factor binding protein 6 ; IBP6; IGF binding protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518929 349.00
Check your price
  • p63 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536069
  • Size 100µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
p51; p40; Tumor protein p73-like; Transformation-related protein 63; Keratinocyte transcription factor KET; Chronic ulcerative stomatitis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536069 Not Orderable Online
Check your price
  • ZFP36 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541450
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
Zinc finger protein 36 homolog; Protein TIS11A; Growth factor-inducible nuclear protein NUP475; G0/G1 switch regulatory protein 24; Zinc finger protein 36; TPA-induced sequence 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541450 309.00
Check your price
  • Phospho-p63 (Ser455) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538380
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
p51; p40; Tumor protein p73-like; Transformation-related protein 63; Keratinocyte transcription factor KET; Chronic ulcerative stomatitis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538380 Not Orderable Online
Check your price
  • Phospho-p63 (Ser395) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA539827
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
p51; p40; Tumor protein p73-like; Transformation-related protein 63; Keratinocyte transcription factor KET; Chronic ulcerative stomatitis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539827 Not Orderable Online
Check your price
  • p63 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate
  • Catalog # PA121739
  • Size 100µg
Applications
Immunocytochemistry (1:300), Immunofluorescence (1:300), Western Blot (1:1000)
Alternate names:
p51; p40; Tumor protein p73-like; Transformation-related protein 63; Keratinocyte transcription factor KET; Chronic ulcerative stomatitis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121739 Not Orderable Online
Check your price
  • IGFBP6 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547940
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IGFBP6 , insulin-like growth factor binding protein 6 , IBP6, IGF binding protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547940 349.00
Check your price
  • FGFR4 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518084
  • Size 100µL
Applications
Immunoprecipitation (Assay-dependent), Western Blot (1:100)
Alternate names:
TKF; JTK2; CD334; MGC20292; Fibroblast growth factor receptor 4; CD334
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518084 425.00
Check your price