1 - 4 of 4 products displayed
  • KHK Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529004
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Hepatic fructokinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529004 349.00
Check your price
  • KHK Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127839
  • Size 50 µg
Applications
Western Blot (1:2000)
Alternate names:
Hepatic fructokinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127839 425.00
Check your price
  • KHK Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA519238
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Hepatic fructokinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA519238 309.00
Check your price
  • KHK Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541992
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Hepatic fructokinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541992 309.00
Check your price