1 - 11 of 11 products displayed
  • Ki-67 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515690
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
KIA; Ki-67; MKI67
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515690 349.00
Check your price
  • Ki-67 Monoclonal Antibody for IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515525
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000)
Alternate names:
KIA; Ki-67; MKI67
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515525 349.00
Check your price
  • Ki-67 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (Free), IHC (P) and Flow
  • Host Rabbit
  • Target Species Bovine, Dog, Deer, Horse, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA519462
  • Size 100 µg
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry-Free Floating (1:500), Immunohistochemistry (Frozen) (1:100-1:1000), Immunohistochemistry (Paraffin) (0.1-10 µg/mL), Western Blot (1 µg/ml)
Alternate names:
RP11-380J17.2, KIA, antigen KI-67; proliferation-related Ki-67 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519462 349.00
Check your price
  • Ki-67 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515484
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
KIA; Ki-67; MKI67
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515484 349.00
Check your price
  • Ki-67 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA516446
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (10-20 µg/mL)
Alternate names:
antigen KI-67, proliferation-related Ki-67 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516446 309.00
Check your price
  • Ki-67 Monoclonal Antibody for IF, ICC, Flow and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701198
  • Size 100 µg
Applications
ChIP assay (1 ul), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500)
Alternate names:
antigen KI-67, proliferation-related Ki-67 antigen, RP11-380J17.2, KIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
701198 303.00
Check your price
  • Ki-67 Oligoclonal Antibody for IF, ICC and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710229
  • Size 100 µg
Applications
ChIP assay (1 ul), Immunocytochemistry (1-3 ug/ml), Immunofluorescence (1-3 ug/ml)
Alternate names:
antigen KI-67, proliferation-related Ki-67 antigen, RP11-380J17.2, KIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
710229 309.00
Check your price
  • Ki-67 Polyclonal Antibody for IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA516785
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:300)
Alternate names:
antigen KI-67, proliferation-related Ki-67 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516785 309.00
Check your price
  • Ki-67 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121520
  • Size 500 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (10-20 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121520 399.00
Check your price
  • NIFK Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA141143
  • Size 100 µg
Applications
Western Blot (0.5-3 µg/ml)
Alternate names:
Nucleolar protein interacting with the FHA domain of pKI-67; Nucleolar phosphoprotein Nopp34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141143 325.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, Qdot® 585 conjugate for Western Blot, ICC, IHC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Qdot® 585
  • Catalog # Q11411MP
  • Size 200 µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunohistochemistry (1:50), Western Blot (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
Q11411MP 313.00
Check your price