1 - 4 of 4 products displayed
  • Phospho-KIDINS220 (Ser918) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14251
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Ankyrin repeat-rich membrane-spanning protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14251 309.00
Check your price
  • KIDINS220 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14229
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Ankyrin repeat-rich membrane-spanning protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14229 309.00
Check your price
  • KIDINS220 Monoclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA190667
  • Size 100µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Ankyrin repeat-rich membrane-spanning protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190667 375.00
Check your price
  • KIDINS220 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522116
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Ankyrin repeat-rich membrane-spanning protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522116 349.00
Check your price