1 - 30 of 30 products displayed
  • p70 S6 Kinase Antibody (Oligoclonal, 15HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710285
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:100-1:1000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710285 304.00
Check your price
  • Creatine Kinase BB Antibody (Monoclonal, BDI937)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191183
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Creatine kinase B chain; B-CK
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191183 399.00
Check your price
  • Phospho-DAPK1 (Ser308) Antibody (Monoclonal, DKPS308)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA124738
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1-2 µg/ml)
Alternate names:
DAPK, DKFZp781I035
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124738 349.00
Check your price
  • p70 S6 Kinase Antibody (Monoclonal, 5G9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517168
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517168 309.00
Check your price
  • p70 S6 Kinase Antibody
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131167
  • Size 100 µg
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
70 kDa ribosomal protein S6 kinase 1; p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131167 349.00
Check your price
  • Adenylate Kinase 1 Antibody (Monoclonal, 3G8-1B11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518367
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Myokinase; Adenylate monophosphate kinase; ATP:AMP phosphotransferase; ATP-AMP transphosphorylase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518367 309.00
Check your price
  • Adenylate Kinase 9 Antibody (Monoclonal, 1H8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523189
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunohistochemistry (PFA fixed) (Assay-Dependent)
Alternate names:
Adenylate kinase domain-containing protein 2; Adenylate kinase domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523189 309.00
Check your price
  • Glycerol kinase 2 Antibody (Monoclonal, 3G4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518967
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
Glycerol kinase, testis specific 2; ATP:glycerol 3-phosphotransferase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518967 309.00
Check your price
  • Creatine Kinase MB Antibody (Monoclonal, 46A1)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # LFMA0233
  • Size 100 µL
Applications
ELISA (Assay Dependent), Western Blot (1:2000)
Alternate names:
M-CK; Creatine kinase M chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0233 309.00
Check your price
  • CaMKII alpha Antibody (Monoclonal, Cba-2)
  • Host Mouse
  • Target Species Fish, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # 137300
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, calcium/calmodulin-dependent protein kinase type II alpha chain, Calci
Catalog # Size Price (AUD) Your Price (AUD) Qty
137300 304.00
Check your price
  • CK1 epsilon Antibody (Monoclonal, AF6C1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517231
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (1 ul/400 ul lysate), Western Blot (1:2000)
Alternate names:
casein kinase 1 epsilon; CK1e; CKIe; RP1-5O6.1; HCKIE; MGC10398 ; CKI-epsilon; OTTHUMP00000063258; OTTHUMP00000063263; casein kinase 1 epsilon; casein kinase I epsilon
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517231 349.00
Check your price
  • Phospho-CaMKII alpha (Thr286) Antibody (Monoclonal, 22B1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1047
  • Size 100 µL
Applications
ELISA (Assay dependent), Flow Cytometry (1µg per 10^6 cells), Immunocytochemistry (1:100), Immunofluorescence (1:50), Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type 2, Phospho-CaMKI
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1047 375.00
Check your price
  • CaMKII beta Antibody (Monoclonal, CB-beta-1)
  • Host Mouse
  • Target Species Fish, Human, Mouse, Sheep, Rat, Sea urchin
  • Conjugate Unconjugated
  • Catalog # 139800
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase II delta, calcium/calmodulin-dependent protein kinase type II delta chain, Calcium/calmodulin-dependent protein kinase type II subunit delta, Ca
Catalog # Size Price (AUD) Your Price (AUD) Qty
139800 304.00
Check your price
  • GRK2 Antibody (Monoclonal, 5D5)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA12013
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (Assay dependent), Western Blot (3-5 ug/ml)
Alternate names:
G-protein-coupled receptor kinase 2; G-protein coupled receptor kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA12013 275.00
Check your price
  • ACVRL1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120176
  • Size 25 µg
Applications
ELISA (0.5-1 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
TGF-B superfamily receptor type I; Activin receptor-like kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120176 349.00
Check your price
  • LKB1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112767
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Renal carcinoma antigen NY-REN-19; Liver kinase B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112767 449.00
Check your price
  • GSK3 beta Antibody
  • Host Chicken
  • Target Species Rat, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA127893
  • Size 50 µg
Applications
ELISA (5 µg/ml), Western Blot (Assay Dependent)
Alternate names:
Serine/threonine-protein kinase GSK3B; Factor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127893 425.00
Check your price
  • CK1 delta Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA128034
  • Size 50 µL
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Tau-protein kinase CSNK1D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128034 349.00
Check your price
  • Aurora B Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Pig, Rat, Bovine
  • Conjugate Unconjugated
  • Catalog # PA186828
  • Size 50 µg
Applications
ELISA (1:2000-1:10,000), Western Blot (1:250-1:1000)
Alternate names:
Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 12; STK-1; Aurora/IPL1-related kinase 2; Aurora- and IPL1-like midbody-associated protein 1; Aurora 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186828 425.00
Check your price
  • PLK1 Antibody (Monoclonal, 35-206)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 377000
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Serine/threonine-protein kinase 13; Polo-like kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
377000 305.00
Check your price
  • DMPK Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 349900
  • Size 50 µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Myotonic dystrophy protein kinase; DMPK; DM-kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
349900 304.00
Check your price
  • SGK2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533019
  • Size 50 ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
Serum/glucocorticoid-regulated kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533019 349.00
Check your price
  • GRK2 Antibody (Monoclonal, 5D5)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 350100Z
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
G-protein-coupled receptor kinase 2; G-protein coupled receptor kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
350100Z 309.00
Check your price
  • CERK Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112768
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Lipid kinase 4; Acylsphingosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112768 449.00
Check your price
  • CERK Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112765
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Lipid kinase 4; Acylsphingosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112765 449.00
Check your price
  • CK2 beta Antibody (Monoclonal, 26C3)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0216
  • Size 100 µL
Applications
ELISA (Assay Dependent), Western Blot (1:2000)
Alternate names:
Protein G5a; Phosvitin
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0216 309.00
Check your price
  • PRKAR2A Antibody (Monoclonal, RS17)
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA120072
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent)
Alternate names:
MGC3606, PKR2, PRKAR2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120072 309.00
Check your price
  • PRKAR2A Antibody (Monoclonal, RS30)
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA120070
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent)
Alternate names:
MGC3606, PKR2, PRKAR2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120070 309.00
Check your price
  • PRKAR2A Antibody (Monoclonal, RS6)
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA120071
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent)
Alternate names:
MGC3606, PKR2, PRKAR2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120071 309.00
Check your price
  • PRKD2 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA123446
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
nPKC-D2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123446 375.00
Check your price