1 - 8 of 8 products displayed
  • Kinesin 5B Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # PA119392
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA119392 275.00
Check your price
  • Kinesin 5B Antibody (Monoclonal, KN-02)
  • Host Mouse
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA119352
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (1:100-1:500)
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119352 275.00
Check your price
  • Kinesin 5B Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521623
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521623 349.00
Check your price
  • Kinesin 5B Antibody (Monoclonal, KN-03)
  • Host Mouse
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA119353
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent)
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119353 225.00
Check your price
  • Kinesin 5B Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1643
  • Size 100 µg
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Immunoprecipitation (5 µg), Western Blot (1:4000-1:6000 )
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1643 375.00
Check your price
  • Kinesin 5B Antibody
  • Host Goat
  • Target Species Dog, Human
  • Conjugate Unconjugated
  • Catalog # PA518063
  • Size 100 ug
Applications
Immunohistochemistry (1-2µg/ml), Western Blot (0.2-1 ug/ml)
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518063 349.00
Check your price
  • Kinesin 5B Antibody
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131243
  • Size 100 µg
Applications
ELISA (1:32,000), Immunohistochemistry (Paraffin) (1-3 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131243 399.00
Check your price
  • Human Kinesin 5B (aa 376-396) Synthetic Peptide
  • Host N/A
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PEP198
  • Size 50 µg
Applications
Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP198 109.00
Check your price